Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 15613918-15614149 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDRT1
Entrez Gene ID
374286
Gene Name
CMT1A duplicated region transcript 1
Gene Aliases
C170RF1, C17ORF1, C17ORF1A, FBXW10B, FBXW10P1, HREP, SM25H2
UniGene
Hs.548021
Gene Chromosome Location
Chr.17: 15565482-15619704 on Build GRCh38
Transcripts
NM_001282540.1 NM_006382.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3932486 c.734C>G p.A245G Chr.17: 15613970-15613970 on Build GRCh38
1324144 c.704C>T p.S235F Chr.17: 15614000-15614000 on Build GRCh38
4129601 c.781C>G p.L261V Chr.17: 15613923-15613923 on Build GRCh38
4000022 c.781C>A p.L261M Chr.17: 15613923-15613923 on Build GRCh38
4671269 c.705C>T p.S235S Chr.17: 15613999-15613999 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374589141 Chr.17: 15614069-15614069 on Build GRCh38
rs779426704 Chr.17: 15613973-15613973 on Build GRCh38
rs902885788 Chr.17: 15614122-15614122 on Build GRCh38
rs564013549 Chr.17: 15614039-15614039 on Build GRCh38
rs60687202 Chr.17: 15614059-15614059 on Build GRCh38
rs753552395 Chr.17: 15613944-15613944 on Build GRCh38
rs765288694 Chr.17: 15613951-15613951 on Build GRCh38
rs774849864 Chr.17: 15614009-15614009 on Build GRCh38
rs139932004 Chr.17: 15614095-15614095 on Build GRCh38
rs771234241 Chr.17: 15614000-15614000 on Build GRCh38
rs575128109 Chr.17: 15613953-15613953 on Build GRCh38
rs368578151 Chr.17: 15614010-15614010 on Build GRCh38
rs950353291 Chr.17: 15614101-15614101 on Build GRCh38
rs190605576 Chr.17: 15614097-15614097 on Build GRCh38
rs117174935 Chr.17: 15614058-15614058 on Build GRCh38
rs761716050 Chr.17: 15614047-15614047 on Build GRCh38
rs3809727 Chr.17: 15613970-15613970 on Build GRCh38
rs56926918 Chr.17: 15614092-15614092 on Build GRCh38
rs765057695 Chr.17: 15614050-15614050 on Build GRCh38
rs377200473 Chr.17: 15613948-15613948 on Build GRCh38
rs746737120 Chr.17: 15613983-15613983 on Build GRCh38
rs113130850 Chr.17: 15613998-15613998 on Build GRCh38
rs772203218 Chr.17: 15614022-15614022 on Build GRCh38
rs560619145 Chr.17: 15614075-15614075 on Build GRCh38
rs752286072 Chr.17: 15614083-15614083 on Build GRCh38
rs922653782 Chr.17: 15614015-15614015 on Build GRCh38
rs1046402019 Chr.17: 15614120-15614120 on Build GRCh38
rs776865248 Chr.17: 15614044-15614044 on Build GRCh38
rs778242833 Chr.17: 15613999-15613999 on Build GRCh38
rs764189913 Chr.17: 15614041-15614041 on Build GRCh38
rs529620815 Chr.17: 15614078-15614078 on Build GRCh38
rs750452819 Chr.17: 15614056-15614056 on Build GRCh38
rs775417718 Chr.17: 15614035-15614035 on Build GRCh38
rs750329974 Chr.17: 15613961-15613961 on Build GRCh38
rs761003984 Chr.17: 15614037-15614037 on Build GRCh38

View Full Product Details