Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 86227224-86227497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf153
Entrez Gene ID
389766
Gene Name
chromosome 9 open reading frame 153
Gene Aliases
bA507D14.1
UniGene
Hs.632073
Gene Chromosome Location
Chr.9: 86220265-86259657 on Build GRCh38
Transcripts
NM_001276366.1 NM_001276367.1 NM_001276368.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3659300 c.275G>A p.R92K Chr.9: 86227364-86227364 on Build GRCh38
5446526 c.292_293insAAATAAAT p.? Chr.9: 86227346-86227347 on Build GRCh38
456181 c.243-2A>T p.? Chr.9: 86227398-86227398 on Build GRCh38
5411418 c.289_290CC>TT p.P97L Chr.9: 86227349-86227350 on Build GRCh38
6616840 c.257A>T p.N86I Chr.9: 86227382-86227382 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs181012219 Chr.9: 86227320-86227320 on Build GRCh38
rs202203722 Chr.9: 86227291-86227291 on Build GRCh38
rs1028843577 Chr.9: 86227271-86227271 on Build GRCh38
rs201401346 Chr.9: 86227314-86227314 on Build GRCh38
rs779943655 Chr.9: 86227374-86227374 on Build GRCh38
rs759041929 Chr.9: 86227399-86227399 on Build GRCh38
rs138074918 Chr.9: 86227315-86227319 on Build GRCh38
rs1022722893 Chr.9: 86227248-86227248 on Build GRCh38
rs768503689 Chr.9: 86227359-86227359 on Build GRCh38
rs780450475 Chr.9: 86227316-86227323 on Build GRCh38
rs201744679 Chr.9: 86227367-86227367 on Build GRCh38
rs140720945 Chr.9: 86227366-86227366 on Build GRCh38
rs751235011 Chr.9: 86227395-86227395 on Build GRCh38
rs1041163937 Chr.9: 86227416-86227416 on Build GRCh38
rs146954955 Chr.9: 86227315-86227316 on Build GRCh38
rs541797695 Chr.9: 86227319-86227320 on Build GRCh38
rs748931055 Chr.9: 86227316-86227331 on Build GRCh38
rs768335029 Chr.9: 86227324-86227325 on Build GRCh38
rs747901061 Chr.9: 86227407-86227408 on Build GRCh38
rs987380335 Chr.9: 86227308-86227308 on Build GRCh38
rs762586195 Chr.9: 86227371-86227371 on Build GRCh38
rs779224955 Chr.9: 86227293-86227293 on Build GRCh38
rs920238168 Chr.9: 86227277-86227277 on Build GRCh38
rs778262594 Chr.9: 86227354-86227355 on Build GRCh38
rs116991392 Chr.9: 86227267-86227267 on Build GRCh38
rs747713773 Chr.9: 86227359-86227362 on Build GRCh38
rs926305274 Chr.9: 86227408-86227408 on Build GRCh38
rs370789582 Chr.9: 86227316-86227319 on Build GRCh38
rs372229263 Chr.9: 86227368-86227368 on Build GRCh38
rs745732180 Chr.9: 86227272-86227272 on Build GRCh38
rs780916694 Chr.9: 86227266-86227266 on Build GRCh38
rs939575166 Chr.9: 86227458-86227458 on Build GRCh38
rs921384936 Chr.9: 86227421-86227421 on Build GRCh38
rs756769274 Chr.9: 86227243-86227243 on Build GRCh38
rs78503535 Chr.9: 86227349-86227349 on Build GRCh38
rs930278786 Chr.9: 86227402-86227402 on Build GRCh38

View Full Product Details