Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 107069680-107069937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEND3
Entrez Gene ID
57673
Gene Name
BEN domain containing 3
Gene Aliases
KIAA1553
UniGene
Hs.418045
Gene Chromosome Location
Chr.6: 107065180-107115187 on Build GRCh38
Transcripts
NM_001080450.2 XM_005267079.3 XM_005267080.3 XM_011536005.2 XM_017011138.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3975710 c.1269C>T p.F423F Chr.6: 107069922-107069922 on Build GRCh38
4628471 c.1363A>G p.T455A Chr.6: 107069828-107069828 on Build GRCh38
3941445 c.1411C>T p.Q471* Chr.6: 107069780-107069780 on Build GRCh38
7347539 c.1366G>T p.E456* Chr.6: 107069825-107069825 on Build GRCh38
5566243 c.1484G>A p.R495H Chr.6: 107069707-107069707 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781992169 Chr.6: 107069806-107069806 on Build GRCh38
rs781947441 Chr.6: 107069769-107069769 on Build GRCh38
rs782738425 Chr.6: 107069790-107069790 on Build GRCh38
rs782465261 Chr.6: 107069837-107069837 on Build GRCh38
rs200244436 Chr.6: 107069851-107069851 on Build GRCh38
rs909648129 Chr.6: 107069854-107069854 on Build GRCh38
rs147116683 Chr.6: 107069852-107069852 on Build GRCh38
rs782447705 Chr.6: 107069754-107069754 on Build GRCh38
rs140372572 Chr.6: 107069853-107069853 on Build GRCh38
rs370526148 Chr.6: 107069718-107069718 on Build GRCh38
rs781837960 Chr.6: 107069836-107069836 on Build GRCh38
rs782365930 Chr.6: 107069875-107069875 on Build GRCh38
rs146427358 Chr.6: 107069884-107069884 on Build GRCh38
rs782063371 Chr.6: 107069827-107069827 on Build GRCh38
rs782117275 Chr.6: 107069917-107069917 on Build GRCh38
rs993192661 Chr.6: 107069826-107069826 on Build GRCh38
rs782316621 Chr.6: 107069857-107069857 on Build GRCh38
rs374180416 Chr.6: 107069731-107069731 on Build GRCh38
rs564584401 Chr.6: 107069727-107069727 on Build GRCh38
rs77387720 Chr.6: 107069715-107069715 on Build GRCh38
rs781888818 Chr.6: 107069823-107069823 on Build GRCh38
rs782406426 Chr.6: 107069768-107069768 on Build GRCh38
rs764064624 Chr.6: 107069913-107069913 on Build GRCh38
rs782093278 Chr.6: 107069905-107069905 on Build GRCh38
rs782052549 Chr.6: 107069780-107069780 on Build GRCh38
rs782252376 Chr.6: 107069869-107069869 on Build GRCh38
rs75435915 Chr.6: 107069751-107069751 on Build GRCh38
rs782037886 Chr.6: 107069710-107069710 on Build GRCh38
rs782587231 Chr.6: 107069865-107069865 on Build GRCh38
rs138792120 Chr.6: 107069886-107069886 on Build GRCh38
rs782612306 Chr.6: 107069757-107069757 on Build GRCh38
rs368969067 Chr.6: 107069776-107069776 on Build GRCh38
rs782542029 Chr.6: 107069744-107069744 on Build GRCh38
rs559431190 Chr.6: 107069714-107069714 on Build GRCh38
rs781991499 Chr.6: 107069766-107069766 on Build GRCh38
rs781921986 Chr.6: 107069900-107069900 on Build GRCh38
rs782588658 Chr.6: 107069760-107069760 on Build GRCh38
rs140588739 Chr.6: 107069843-107069843 on Build GRCh38
rs775818889 Chr.6: 107069799-107069799 on Build GRCh38
rs1034647476 Chr.6: 107069809-107069809 on Build GRCh38
rs782303650 Chr.6: 107069767-107069767 on Build GRCh38
rs782323136 Chr.6: 107069885-107069885 on Build GRCh38
rs782188646 Chr.6: 107069753-107069753 on Build GRCh38
rs41292438 Chr.6: 107069880-107069880 on Build GRCh38
rs959977844 Chr.6: 107069816-107069816 on Build GRCh38
rs782196758 Chr.6: 107069856-107069856 on Build GRCh38
rs782805647 Chr.6: 107069814-107069814 on Build GRCh38
rs190547697 Chr.6: 107069707-107069707 on Build GRCh38
rs782214420 Chr.6: 107069802-107069802 on Build GRCh38
rs782690060 Chr.6: 107069828-107069828 on Build GRCh38
rs782112705 Chr.6: 107069808-107069808 on Build GRCh38

View Full Product Details