Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26951620-26951879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDF1
Entrez Gene ID
126695
Gene Name
keratinocyte differentiation factor 1
Gene Aliases
C1orf172
UniGene
Hs.188881
Gene Chromosome Location
Chr.1: 26949556-26960484 on Build GRCh38
Transcripts
NM_152365.2 XM_005245735.2 XM_011540622.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5583331 c.502G>A p.G168S Chr.1: 26951879-26951879 on Build GRCh38
1193002 c.521_522delAC p.Y174fs*9 Chr.1: 26951859-26951860 on Build GRCh38
907804 c.632G>A p.R211H Chr.1: 26951749-26951749 on Build GRCh38
3487622 c.507C>T p.I169I Chr.1: 26951874-26951874 on Build GRCh38
907803 c.682G>A p.E228K Chr.1: 26951699-26951699 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149743404 Chr.1: 26951812-26951812 on Build GRCh38
rs759220131 Chr.1: 26951774-26951775 on Build GRCh38
rs369086580 Chr.1: 26951706-26951706 on Build GRCh38
rs766306128 Chr.1: 26951778-26951778 on Build GRCh38
rs746835693 Chr.1: 26951714-26951714 on Build GRCh38
rs376036605 Chr.1: 26951795-26951795 on Build GRCh38
rs754983927 Chr.1: 26951856-26951856 on Build GRCh38
rs780718896 Chr.1: 26951721-26951721 on Build GRCh38
rs777367914 Chr.1: 26951794-26951794 on Build GRCh38
rs766818287 Chr.1: 26951657-26951657 on Build GRCh38
rs1018990840 Chr.1: 26951824-26951824 on Build GRCh38
rs781107975 Chr.1: 26951789-26951789 on Build GRCh38
rs754712166 Chr.1: 26951786-26951786 on Build GRCh38
rs761892290 Chr.1: 26951834-26951834 on Build GRCh38
rs924418483 Chr.1: 26951859-26951859 on Build GRCh38
rs775975243 Chr.1: 26951820-26951820 on Build GRCh38
rs755838886 Chr.1: 26951793-26951793 on Build GRCh38
rs758381938 Chr.1: 26951700-26951700 on Build GRCh38
rs886914997 Chr.1: 26951642-26951642 on Build GRCh38
rs748813446 Chr.1: 26951801-26951801 on Build GRCh38
rs771990259 Chr.1: 26951731-26951731 on Build GRCh38
rs374964459 Chr.1: 26951698-26951698 on Build GRCh38
rs142040426 Chr.1: 26951829-26951829 on Build GRCh38
rs199575697 Chr.1: 26951830-26951830 on Build GRCh38
rs772927829 Chr.1: 26951750-26951750 on Build GRCh38
rs200494012 Chr.1: 26951742-26951742 on Build GRCh38
rs767193569 Chr.1: 26951857-26951857 on Build GRCh38
rs746444382 Chr.1: 26951813-26951813 on Build GRCh38
rs371743170 Chr.1: 26951677-26951677 on Build GRCh38
rs774444521 Chr.1: 26951658-26951658 on Build GRCh38
rs887047479 Chr.1: 26951848-26951848 on Build GRCh38
rs761001620 Chr.1: 26951749-26951749 on Build GRCh38
rs767411580 Chr.1: 26951800-26951801 on Build GRCh38
rs761634533 Chr.1: 26951675-26951675 on Build GRCh38
rs751293758 Chr.1: 26951785-26951785 on Build GRCh38
rs758651584 Chr.1: 26951671-26951671 on Build GRCh38
rs756134971 Chr.1: 26951687-26951687 on Build GRCh38
rs772817105 Chr.1: 26951733-26951733 on Build GRCh38
rs372588334 Chr.1: 26951745-26951745 on Build GRCh38
rs374676574 Chr.1: 26951759-26951759 on Build GRCh38
rs779779172 Chr.1: 26951702-26951702 on Build GRCh38
rs141390568 Chr.1: 26951724-26951724 on Build GRCh38
rs572491048 Chr.1: 26951715-26951715 on Build GRCh38
rs139585050 Chr.1: 26951736-26951736 on Build GRCh38
rs767639195 Chr.1: 26951676-26951676 on Build GRCh38
rs34291506 Chr.1: 26951815-26951815 on Build GRCh38
rs763620227 Chr.1: 26951649-26951649 on Build GRCh38
rs747723043 Chr.1: 26951792-26951792 on Build GRCh38
rs370198466 Chr.1: 26951802-26951802 on Build GRCh38
rs751490034 Chr.1: 26951852-26951852 on Build GRCh38
rs771599585 Chr.1: 26951741-26951741 on Build GRCh38
rs777979399 Chr.1: 26951690-26951690 on Build GRCh38
rs145994096 Chr.1: 26951768-26951768 on Build GRCh38
rs765356214 Chr.1: 26951851-26951851 on Build GRCh38
rs894118968 Chr.1: 26951716-26951716 on Build GRCh38
rs767984255 Chr.1: 26951746-26951746 on Build GRCh38

View Full Product Details