Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 556978-557219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMNTD2
Entrez Gene ID
256329
Gene Name
lamin tail domain containing 2
Gene Aliases
C11orf35
UniGene
Hs.669395
Gene Chromosome Location
Chr.11: 554850-560780 on Build GRCh38
Transcripts
NM_173573.2 XM_011519965.2 XM_011519967.2 XM_017017476.1 XM_017017477.1 XM_017017478.1 XM_017017479.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3967597 c.793G>A p.V265M Chr.11: 557018-557018 on Build GRCh38
4195204 c.815G>A p.C272Y Chr.11: 556996-556996 on Build GRCh38
5768993 c.794T>C p.V265A Chr.11: 557017-557017 on Build GRCh38
1197899 c.722G>A p.R241Q Chr.11: 557089-557089 on Build GRCh38
4910905 c.778C>T p.R260W Chr.11: 557033-557033 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772929924 Chr.11: 557098-557098 on Build GRCh38
rs752136144 Chr.11: 557122-557122 on Build GRCh38
rs774150499 Chr.11: 557019-557019 on Build GRCh38
rs975880134 Chr.11: 557162-557162 on Build GRCh38
rs145512090 Chr.11: 557048-557048 on Build GRCh38
rs775626815 Chr.11: 557112-557112 on Build GRCh38
rs565620275 Chr.11: 557154-557154 on Build GRCh38
rs995362075 Chr.11: 557068-557068 on Build GRCh38
rs984556558 Chr.11: 557152-557152 on Build GRCh38
rs750781882 Chr.11: 557057-557057 on Build GRCh38
rs768212852 Chr.11: 557078-557078 on Build GRCh38
rs377645123 Chr.11: 557118-557118 on Build GRCh38
rs768041791 Chr.11: 557138-557138 on Build GRCh38
rs147297012 Chr.11: 557090-557090 on Build GRCh38
rs747997197 Chr.11: 557089-557089 on Build GRCh38
rs756542492 Chr.11: 557147-557147 on Build GRCh38
rs771717290 Chr.11: 557022-557022 on Build GRCh38
rs1036573362 Chr.11: 557030-557030 on Build GRCh38
rs531119609 Chr.11: 557173-557173 on Build GRCh38
rs763036858 Chr.11: 557115-557115 on Build GRCh38
rs367796723 Chr.11: 557075-557075 on Build GRCh38
rs147157097 Chr.11: 557033-557033 on Build GRCh38
rs140424352 Chr.11: 557036-557036 on Build GRCh38
rs749638605 Chr.11: 557066-557067 on Build GRCh38
rs755875504 Chr.11: 557063-557063 on Build GRCh38
rs549885966 Chr.11: 557155-557155 on Build GRCh38
rs768772255 Chr.11: 557018-557018 on Build GRCh38
rs913079216 Chr.11: 557187-557187 on Build GRCh38
rs748784797 Chr.11: 557014-557014 on Build GRCh38
rs945095431 Chr.11: 557179-557179 on Build GRCh38
rs780909506 Chr.11: 557000-557000 on Build GRCh38
rs745459880 Chr.11: 557001-557001 on Build GRCh38
rs769056869 Chr.11: 557066-557075 on Build GRCh38
rs775176357 Chr.11: 557013-557013 on Build GRCh38
rs750875632 Chr.11: 557146-557146 on Build GRCh38
rs374225356 Chr.11: 557037-557037 on Build GRCh38
rs779839267 Chr.11: 557066-557066 on Build GRCh38
rs576909548 Chr.11: 557007-557007 on Build GRCh38
rs764028077 Chr.11: 557119-557119 on Build GRCh38
rs778933757 Chr.11: 557087-557087 on Build GRCh38
rs771401663 Chr.11: 557179-557180 on Build GRCh38
rs370070902 Chr.11: 557100-557100 on Build GRCh38
rs776944817 Chr.11: 557032-557032 on Build GRCh38
rs988601323 Chr.11: 557185-557185 on Build GRCh38
rs374569827 Chr.11: 557108-557108 on Build GRCh38
rs770071581 Chr.11: 557109-557109 on Build GRCh38

View Full Product Details