Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 74634866-74635121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENC1
Entrez Gene ID
8507
Gene Name
ectodermal-neural cortex 1
Gene Aliases
CCL28, ENC-1, KLHL35, KLHL37, NRPB, PIG10, TP53I10
UniGene
Hs.104925
Gene Chromosome Location
Chr.5: 74627406-74641424 on Build GRCh38
Transcripts
NM_001256574.1 NM_001256575.1 NM_001256576.1 NM_003633.3 NR_046318.1 XM_011543696.2 XM_011543697.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7411689 c.1521T>C p.S507S Chr.5: 74634965-74634965 on Build GRCh38
6513105 c.1517G>T p.C506F Chr.5: 74634969-74634969 on Build GRCh38
5481999 c.1585A>G p.M529V Chr.5: 74634901-74634901 on Build GRCh38
330198 c.1544C>A p.T515N Chr.5: 74634942-74634942 on Build GRCh38
1438536 c.1459G>A p.A487T Chr.5: 74635027-74635027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751291467 Chr.5: 74634891-74634891 on Build GRCh38
rs748480145 Chr.5: 74634986-74634986 on Build GRCh38
rs755622645 Chr.5: 74634969-74634969 on Build GRCh38
rs888489840 Chr.5: 74635061-74635061 on Build GRCh38
rs754526395 Chr.5: 74634981-74634981 on Build GRCh38
rs1049448492 Chr.5: 74634993-74634993 on Build GRCh38
rs201897189 Chr.5: 74635064-74635064 on Build GRCh38
rs139465896 Chr.5: 74634944-74634944 on Build GRCh38
rs747333081 Chr.5: 74634975-74634975 on Build GRCh38
rs11747630 Chr.5: 74635068-74635068 on Build GRCh38
rs771037231 Chr.5: 74635066-74635066 on Build GRCh38
rs993452761 Chr.5: 74634962-74634962 on Build GRCh38
rs749403473 Chr.5: 74634956-74634963 on Build GRCh38
rs1039068030 Chr.5: 74634989-74634989 on Build GRCh38
rs779337258 Chr.5: 74634984-74634984 on Build GRCh38
rs748959019 Chr.5: 74634979-74634979 on Build GRCh38
rs746631875 Chr.5: 74635091-74635091 on Build GRCh38
rs759810274 Chr.5: 74635090-74635090 on Build GRCh38
rs776773829 Chr.5: 74635081-74635081 on Build GRCh38
rs374922468 Chr.5: 74634887-74634887 on Build GRCh38
rs300239 Chr.5: 74634926-74634926 on Build GRCh38
rs773833247 Chr.5: 74635028-74635028 on Build GRCh38
rs772724462 Chr.5: 74634990-74634990 on Build GRCh38
rs779643005 Chr.5: 74634971-74634971 on Build GRCh38
rs900505527 Chr.5: 74634991-74634991 on Build GRCh38
rs146381927 Chr.5: 74634938-74634938 on Build GRCh38
rs149679487 Chr.5: 74634972-74634972 on Build GRCh38

View Full Product Details