Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41124722-41124983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2F1
Entrez Gene ID
1572
Gene Name
cytochrome P450 family 2 subfamily F member 1
Gene Aliases
C2F1, CYP2F, CYPIIF1
UniGene
Hs.558318
Gene Chromosome Location
Chr.19: 41114292-41128381 on Build GRCh38
Transcripts
NM_000774.4 NR_135528.1 XM_011526551.2 XM_011526552.2 XM_011526553.2 XM_011526554.2 XM_011526555.2 XM_017026384.1 XM_017026385.1 XM_017026387.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4078419 c.1027C>T p.R343C Chr.19: 41124781-41124781 on Build GRCh38
3404263 c.1134C>T p.R378R Chr.19: 41124888-41124888 on Build GRCh38
4078418 c.984C>T p.I328I Chr.19: 41124738-41124738 on Build GRCh38
3404262 c.1017G>A p.A339A Chr.19: 41124771-41124771 on Build GRCh38
4829306 c.1006C>T p.R336W Chr.19: 41124760-41124760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747567361 Chr.19: 41124840-41124840 on Build GRCh38
rs139008556 Chr.19: 41124806-41124806 on Build GRCh38
rs775168328 Chr.19: 41124935-41124935 on Build GRCh38
rs758023108 Chr.19: 41124802-41124816 on Build GRCh38
rs541072855 Chr.19: 41124923-41124923 on Build GRCh38
rs766004612 Chr.19: 41124759-41124759 on Build GRCh38
rs777720181 Chr.19: 41124769-41124769 on Build GRCh38
rs764372990 Chr.19: 41124879-41124879 on Build GRCh38
rs186961295 Chr.19: 41124940-41124940 on Build GRCh38
rs757331747 Chr.19: 41124827-41124827 on Build GRCh38
rs368079396 Chr.19: 41124863-41124863 on Build GRCh38
rs772617651 Chr.19: 41124789-41124789 on Build GRCh38
rs771675537 Chr.19: 41124927-41124927 on Build GRCh38
rs780407249 Chr.19: 41124838-41124838 on Build GRCh38
rs138507242 Chr.19: 41124878-41124878 on Build GRCh38
rs950111044 Chr.19: 41124784-41124784 on Build GRCh38
rs373232027 Chr.19: 41124782-41124782 on Build GRCh38
rs552985767 Chr.19: 41124757-41124757 on Build GRCh38
rs979206615 Chr.19: 41124762-41124762 on Build GRCh38
rs761659772 Chr.19: 41124950-41124950 on Build GRCh38
rs774133304 Chr.19: 41124924-41124924 on Build GRCh38
rs761170962 Chr.19: 41124785-41124785 on Build GRCh38
rs779057628 Chr.19: 41124832-41124832 on Build GRCh38
rs371341517 Chr.19: 41124857-41124857 on Build GRCh38
rs368705802 Chr.19: 41124906-41124906 on Build GRCh38
rs762465626 Chr.19: 41124855-41124855 on Build GRCh38
rs150197628 Chr.19: 41124805-41124805 on Build GRCh38
rs764600264 Chr.19: 41124786-41124786 on Build GRCh38
rs756135831 Chr.19: 41124813-41124813 on Build GRCh38
rs376080668 Chr.19: 41124760-41124760 on Build GRCh38
rs369493486 Chr.19: 41124781-41124781 on Build GRCh38
rs199974498 Chr.19: 41124826-41124826 on Build GRCh38
rs372302293 Chr.19: 41124911-41124911 on Build GRCh38
rs749333476 Chr.19: 41124770-41124770 on Build GRCh38
rs151064205 Chr.19: 41124750-41124750 on Build GRCh38
rs777087761 Chr.19: 41124844-41124844 on Build GRCh38
rs746081050 Chr.19: 41124833-41124833 on Build GRCh38
rs748550297 Chr.19: 41124915-41124915 on Build GRCh38
rs544175992 Chr.19: 41124843-41124843 on Build GRCh38
rs763688043 Chr.19: 41124906-41124906 on Build GRCh38
rs386809335 Chr.19: 41124805-41124806 on Build GRCh38
rs754170770 Chr.19: 41124754-41124755 on Build GRCh38
rs1045994414 Chr.19: 41124795-41124795 on Build GRCh38
rs141890168 Chr.19: 41124858-41124858 on Build GRCh38
rs115159306 Chr.19: 41124864-41124864 on Build GRCh38
rs760477463 Chr.19: 41124939-41124939 on Build GRCh38
rs775948155 Chr.19: 41124739-41124739 on Build GRCh38
rs759309456 Chr.19: 41124926-41124926 on Build GRCh38
rs771083555 Chr.19: 41124771-41124771 on Build GRCh38
rs756329929 Chr.19: 41124768-41124768 on Build GRCh38
rs758453484 Chr.19: 41124886-41124886 on Build GRCh38
rs371125523 Chr.19: 41124783-41124783 on Build GRCh38
rs577128494 Chr.19: 41124882-41124882 on Build GRCh38
rs764719015 Chr.19: 41124754-41124754 on Build GRCh38
rs762483034 Chr.19: 41124792-41124792 on Build GRCh38
rs772211507 Chr.19: 41124835-41124835 on Build GRCh38
rs749878742 Chr.19: 41124756-41124756 on Build GRCh38
rs780150882 Chr.19: 41124887-41124887 on Build GRCh38
rs879148271 Chr.19: 41124854-41124854 on Build GRCh38
rs777718696 Chr.19: 41124819-41124819 on Build GRCh38
rs753505427 Chr.19: 41124944-41124944 on Build GRCh38
rs73545063 Chr.19: 41124804-41124804 on Build GRCh38
rs925126589 Chr.19: 41124766-41124766 on Build GRCh38
rs568085061 Chr.19: 41124767-41124767 on Build GRCh38
rs765985687 Chr.19: 41124794-41124794 on Build GRCh38
rs367923974 Chr.19: 41124865-41124865 on Build GRCh38
rs752474586 Chr.19: 41124807-41124807 on Build GRCh38
rs368904004 Chr.19: 41124955-41124955 on Build GRCh38
rs112698282 Chr.19: 41124960-41124960 on Build GRCh38
rs376838978 Chr.19: 41124951-41124951 on Build GRCh38
rs774594954 Chr.19: 41124777-41124777 on Build GRCh38
rs202164777 Chr.19: 41124860-41124860 on Build GRCh38

View Full Product Details