Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116379282-116379519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC172
Entrez Gene ID
374355
Gene Name
coiled-coil domain containing 172
Gene Aliases
C10orf96
UniGene
Hs.233407
Gene Chromosome Location
Chr.10: 116324097-116380029 on Build GRCh38
Transcripts
NM_198515.2 XM_006717820.3 XM_017016206.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5919819 c.742-8C>A p.? Chr.10: 116379315-116379315 on Build GRCh38
915139 c.760C>A p.L254I Chr.10: 116379341-116379341 on Build GRCh38
4558397 c.766G>A p.E256K Chr.10: 116379347-116379347 on Build GRCh38
3375394 c.742-4T>G p.? Chr.10: 116379319-116379319 on Build GRCh38
135984 c.770G>A p.S257N Chr.10: 116379351-116379351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750786833 Chr.10: 116379425-116379425 on Build GRCh38
rs763140561 Chr.10: 116379476-116379477 on Build GRCh38
rs773561597 Chr.10: 116379470-116379471 on Build GRCh38
rs943387815 Chr.10: 116379367-116379367 on Build GRCh38
rs763901835 Chr.10: 116379394-116379394 on Build GRCh38
rs376129168 Chr.10: 116379351-116379351 on Build GRCh38
rs759699313 Chr.10: 116379375-116379375 on Build GRCh38
rs878996833 Chr.10: 116379449-116379449 on Build GRCh38
rs1025170227 Chr.10: 116379446-116379446 on Build GRCh38
rs150979898 Chr.10: 116379326-116379326 on Build GRCh38
rs141092544 Chr.10: 116379363-116379364 on Build GRCh38
rs534184832 Chr.10: 116379442-116379442 on Build GRCh38
rs112371131 Chr.10: 116379463-116379463 on Build GRCh38
rs531960965 Chr.10: 116379396-116379396 on Build GRCh38
rs755754206 Chr.10: 116379406-116379406 on Build GRCh38
rs771262905 Chr.10: 116379316-116379316 on Build GRCh38
rs765501337 Chr.10: 116379451-116379451 on Build GRCh38
rs192670502 Chr.10: 116379441-116379441 on Build GRCh38
rs767540479 Chr.10: 116379398-116379398 on Build GRCh38
rs146631810 Chr.10: 116379443-116379443 on Build GRCh38
rs773566322 Chr.10: 116379324-116379324 on Build GRCh38
rs1053517168 Chr.10: 116379430-116379430 on Build GRCh38
rs370219167 Chr.10: 116379470-116379470 on Build GRCh38
rs12254317 Chr.10: 116379427-116379427 on Build GRCh38
rs754067202 Chr.10: 116379357-116379357 on Build GRCh38
rs766439410 Chr.10: 116379339-116379339 on Build GRCh38

View Full Product Details