Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 110308049-110308282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL4A2
Entrez Gene ID
1284
Gene Name
collagen type IV alpha 2
Gene Aliases
ICH, POREN2
UniGene
Hs.508716
Gene Chromosome Location
Chr.13: 110307284-110513027 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6633180 c.70G>A p.V24M Chr.13: 110308094-110308094 on Build GRCh38
6633172 c.91G>A p.V31I Chr.13: 110308115-110308115 on Build GRCh38
5501941 c.45-1G>C p.? Chr.13: 110308068-110308068 on Build GRCh38
1248857 c.98C>T p.A33V Chr.13: 110308122-110308122 on Build GRCh38
4821216 c.81C>T p.L27L Chr.13: 110308105-110308105 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs927389346 Chr.13: 110308099-110308099 on Build GRCh38
rs912954865 Chr.13: 110308221-110308221 on Build GRCh38
rs899624739 Chr.13: 110308248-110308248 on Build GRCh38
rs775319592 Chr.13: 110308114-110308114 on Build GRCh38
rs200430407 Chr.13: 110308073-110308073 on Build GRCh38
rs202148198 Chr.13: 110308153-110308153 on Build GRCh38
rs766845623 Chr.13: 110308169-110308169 on Build GRCh38
rs748996226 Chr.13: 110308079-110308079 on Build GRCh38
rs745779326 Chr.13: 110308097-110308097 on Build GRCh38
rs1017916068 Chr.13: 110308237-110308237 on Build GRCh38
rs200523295 Chr.13: 110308139-110308139 on Build GRCh38
rs539903754 Chr.13: 110308228-110308228 on Build GRCh38
rs762167800 Chr.13: 110308152-110308152 on Build GRCh38
rs760739260 Chr.13: 110308116-110308116 on Build GRCh38
rs770707066 Chr.13: 110308084-110308084 on Build GRCh38
rs750767559 Chr.13: 110308160-110308160 on Build GRCh38
rs1043009281 Chr.13: 110308091-110308091 on Build GRCh38
rs947169071 Chr.13: 110308223-110308223 on Build GRCh38
rs888669203 Chr.13: 110308222-110308222 on Build GRCh38
rs755584763 Chr.13: 110308075-110308075 on Build GRCh38
rs916555766 Chr.13: 110308123-110308123 on Build GRCh38
rs200428476 Chr.13: 110308127-110308127 on Build GRCh38
rs12854654 Chr.13: 110308162-110308162 on Build GRCh38
rs774171520 Chr.13: 110308093-110308093 on Build GRCh38
rs935490324 Chr.13: 110308199-110308199 on Build GRCh38
rs777427868 Chr.13: 110308078-110308078 on Build GRCh38
rs895976079 Chr.13: 110308149-110308149 on Build GRCh38
rs367984136 Chr.13: 110308113-110308113 on Build GRCh38
rs769235344 Chr.13: 110308219-110308220 on Build GRCh38

View Full Product Details