Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26467608-26467835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTN2A1
Entrez Gene ID
11120
Gene Name
butyrophilin subfamily 2 member A1
Gene Aliases
BK14H9.1, BT2.1, BTF1, BTN2.1, DJ3E1.1
UniGene
Hs.159028
Gene Chromosome Location
Chr.6: 26457904-26476621 on Build GRCh38
Transcripts
NM_001197233.2 NM_001197234.2 NM_007049.4 NM_078476.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6308637 c.998C>G p.S333* Chr.6: 26467734-26467734 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779168968 Chr.6: 26467685-26467685 on Build GRCh38
rs749202987 Chr.6: 26467739-26467739 on Build GRCh38
rs528741227 Chr.6: 26467639-26467639 on Build GRCh38
rs772395129 Chr.6: 26467719-26467719 on Build GRCh38
rs774120258 Chr.6: 26467747-26467747 on Build GRCh38
rs768685574 Chr.6: 26467740-26467740 on Build GRCh38
rs551768698 Chr.6: 26467726-26467726 on Build GRCh38
rs775001415 Chr.6: 26467736-26467736 on Build GRCh38
rs375412477 Chr.6: 26467805-26467805 on Build GRCh38
rs748595148 Chr.6: 26467707-26467707 on Build GRCh38
rs201625303 Chr.6: 26467749-26467749 on Build GRCh38
rs999627126 Chr.6: 26467766-26467766 on Build GRCh38
rs767406875 Chr.6: 26467750-26467750 on Build GRCh38
rs769310369 Chr.6: 26467734-26467734 on Build GRCh38
rs765813924 Chr.6: 26467788-26467788 on Build GRCh38
rs985586966 Chr.6: 26467720-26467720 on Build GRCh38
rs918554327 Chr.6: 26467767-26467767 on Build GRCh38
rs930253145 Chr.6: 26467795-26467795 on Build GRCh38
rs114905060 Chr.6: 26467759-26467759 on Build GRCh38
rs895139248 Chr.6: 26467704-26467704 on Build GRCh38
rs544256355 Chr.6: 26467806-26467806 on Build GRCh38
rs754991278 Chr.6: 26467770-26467770 on Build GRCh38
rs753907629 Chr.6: 26467768-26467768 on Build GRCh38
rs766598823 Chr.6: 26467765-26467765 on Build GRCh38
rs767924484 Chr.6: 26467728-26467728 on Build GRCh38
rs559999076 Chr.6: 26467638-26467638 on Build GRCh38
rs780893691 Chr.6: 26467724-26467724 on Build GRCh38

View Full Product Details