Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 38698451-38698688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf53
Entrez Gene ID
400359
Gene Name
chromosome 15 open reading frame 53
Gene Aliases
-
UniGene
Hs.448785
Gene Chromosome Location
Chr.15: 38696598-38700038 on Build GRCh38
Transcripts
NM_207444.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4441445 c.493C>A p.L165I Chr.15: 38698498-38698498 on Build GRCh38
3886876 c.502C>T p.P168S Chr.15: 38698507-38698507 on Build GRCh38
4497038 c.488C>T p.S163F Chr.15: 38698493-38698493 on Build GRCh38
1749103 c.509G>A p.G170E Chr.15: 38698514-38698514 on Build GRCh38
6425610 c.458G>A p.G153E Chr.15: 38698463-38698463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775352402 Chr.15: 38698510-38698510 on Build GRCh38
rs747710070 Chr.15: 38698569-38698569 on Build GRCh38
rs201190868 Chr.15: 38698591-38698591 on Build GRCh38
rs368185290 Chr.15: 38698481-38698481 on Build GRCh38
rs781452269 Chr.15: 38698506-38698506 on Build GRCh38
rs755040652 Chr.15: 38698501-38698501 on Build GRCh38
rs566107297 Chr.15: 38698577-38698577 on Build GRCh38
rs746681785 Chr.15: 38698539-38698539 on Build GRCh38
rs771386697 Chr.15: 38698529-38698529 on Build GRCh38
rs201615193 Chr.15: 38698576-38698576 on Build GRCh38
rs753206331 Chr.15: 38698507-38698507 on Build GRCh38
rs1033497469 Chr.15: 38698566-38698566 on Build GRCh38
rs770055341 Chr.15: 38698487-38698487 on Build GRCh38
rs779516896 Chr.15: 38698535-38698535 on Build GRCh38
rs978784707 Chr.15: 38698559-38698559 on Build GRCh38
rs756667200 Chr.15: 38698511-38698511 on Build GRCh38
rs1020511076 Chr.15: 38698502-38698502 on Build GRCh38
rs941497437 Chr.15: 38698629-38698629 on Build GRCh38
rs533074824 Chr.15: 38698590-38698590 on Build GRCh38
rs12595568 Chr.15: 38698602-38698602 on Build GRCh38
rs1037480192 Chr.15: 38698637-38698637 on Build GRCh38
rs927417055 Chr.15: 38698619-38698619 on Build GRCh38
rs762869821 Chr.15: 38698478-38698478 on Build GRCh38
rs775862464 Chr.15: 38698493-38698493 on Build GRCh38
rs768118261 Chr.15: 38698547-38698547 on Build GRCh38
rs778464179 Chr.15: 38698519-38698519 on Build GRCh38
rs7172816 Chr.15: 38698568-38698568 on Build GRCh38
rs749612475 Chr.15: 38698524-38698524 on Build GRCh38
rs964780906 Chr.15: 38698509-38698509 on Build GRCh38

View Full Product Details