Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 168308736-168308979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DACT2
Entrez Gene ID
168002
Gene Name
dishevelled binding antagonist of beta catenin 2
Gene Aliases
C6orf116, DAPPER2, DPR2, bA503C24.7
UniGene
Hs.673059
Gene Chromosome Location
Chr.6: 168292830-168319754 on Build GRCh38
Transcripts
NM_001286350.1 NM_001286351.1 NM_214462.4 NR_104425.1 XM_011535507.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5016950 c.789C>A p.D263E Chr.6: 168308968-168308968 on Build GRCh38
5662022 c.787G>T p.D263Y Chr.6: 168308970-168308970 on Build GRCh38
5369208 c.895G>A p.G299S Chr.6: 168308862-168308862 on Build GRCh38
5737845 c.980C>T p.P327L Chr.6: 168308777-168308777 on Build GRCh38
4763473 c.829A>G p.S277G Chr.6: 168308928-168308928 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868612517 Chr.6: 168308796-168308796 on Build GRCh38
rs765952509 Chr.6: 168308959-168308959 on Build GRCh38
rs895766632 Chr.6: 168308809-168308809 on Build GRCh38
rs35627974 Chr.6: 168308848-168308848 on Build GRCh38
rs777886087 Chr.6: 168308790-168308790 on Build GRCh38
rs570102629 Chr.6: 168308795-168308795 on Build GRCh38
rs918776019 Chr.6: 168308757-168308757 on Build GRCh38
rs747216419 Chr.6: 168308794-168308794 on Build GRCh38
rs754693657 Chr.6: 168308851-168308851 on Build GRCh38
rs559934089 Chr.6: 168308774-168308774 on Build GRCh38
rs532586396 Chr.6: 168308755-168308755 on Build GRCh38
rs778812998 Chr.6: 168308913-168308913 on Build GRCh38
rs768553866 Chr.6: 168308893-168308893 on Build GRCh38
rs867440507 Chr.6: 168308762-168308762 on Build GRCh38
rs749030019 Chr.6: 168308872-168308872 on Build GRCh38
rs531256391 Chr.6: 168308842-168308842 on Build GRCh38
rs541416019 Chr.6: 168308756-168308756 on Build GRCh38
rs1000054867 Chr.6: 168308852-168308852 on Build GRCh38
rs1042000197 Chr.6: 168308827-168308827 on Build GRCh38
rs377756732 Chr.6: 168308943-168308943 on Build GRCh38
rs184039901 Chr.6: 168308953-168308953 on Build GRCh38
rs868810192 Chr.6: 168308942-168308942 on Build GRCh38
rs905245186 Chr.6: 168308806-168308806 on Build GRCh38
rs1028828205 Chr.6: 168308861-168308861 on Build GRCh38
rs935216597 Chr.6: 168308763-168308763 on Build GRCh38
rs753191558 Chr.6: 168308787-168308787 on Build GRCh38
rs1045582968 Chr.6: 168308802-168308802 on Build GRCh38
rs552416999 Chr.6: 168308935-168308935 on Build GRCh38
rs757460539 Chr.6: 168308805-168308805 on Build GRCh38
rs530127614 Chr.6: 168308788-168308788 on Build GRCh38
rs774495453 Chr.6: 168308914-168308914 on Build GRCh38
rs117046907 Chr.6: 168308954-168308954 on Build GRCh38
rs569420736 Chr.6: 168308917-168308917 on Build GRCh38
rs937417037 Chr.6: 168308800-168308800 on Build GRCh38
rs939366648 Chr.6: 168308823-168308823 on Build GRCh38
rs201471118 Chr.6: 168308824-168308824 on Build GRCh38
rs748077283 Chr.6: 168308916-168308916 on Build GRCh38
rs765675691 Chr.6: 168308776-168308776 on Build GRCh38
rs548772905 Chr.6: 168308792-168308792 on Build GRCh38
rs1026339645 Chr.6: 168308936-168308936 on Build GRCh38
rs748084264 Chr.6: 168308915-168308915 on Build GRCh38
rs887363931 Chr.6: 168308901-168308901 on Build GRCh38
rs1018630393 Chr.6: 168308887-168308887 on Build GRCh38
rs1008617939 Chr.6: 168308877-168308877 on Build GRCh38

View Full Product Details