Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136845573-136845806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf172
Entrez Gene ID
389813
Gene Name
chromosome 9 open reading frame 172
Gene Aliases
-
UniGene
Hs.495485
Gene Chromosome Location
Chr.9: 136844415-136847345 on Build GRCh38
Transcripts
NM_001080482.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4423744 c.1364_1365insG p.P457fs*81 Chr.9: 136845778-136845779 on Build GRCh38
6837596 c.1166C>G p.A389G Chr.9: 136845580-136845580 on Build GRCh38
3655859 c.1384C>T p.P462S Chr.9: 136845798-136845798 on Build GRCh38
6616872 c.1263C>T p.D421D Chr.9: 136845677-136845677 on Build GRCh38
5620400 c.1225T>C p.Y409H Chr.9: 136845639-136845639 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774248919 Chr.9: 136845720-136845720 on Build GRCh38
rs761015315 Chr.9: 136845745-136845745 on Build GRCh38
rs565217311 Chr.9: 136845722-136845722 on Build GRCh38
rs777616846 Chr.9: 136845699-136845699 on Build GRCh38
rs750767950 Chr.9: 136845643-136845643 on Build GRCh38
rs772481691 Chr.9: 136845773-136845773 on Build GRCh38
rs575731810 Chr.9: 136845743-136845743 on Build GRCh38
rs958064310 Chr.9: 136845675-136845675 on Build GRCh38
rs766595592 Chr.9: 136845746-136845746 on Build GRCh38
rs754235633 Chr.9: 136845767-136845767 on Build GRCh38
rs771800176 Chr.9: 136845696-136845696 on Build GRCh38
rs746769333 Chr.9: 136845704-136845704 on Build GRCh38
rs752231389 Chr.9: 136845778-136845778 on Build GRCh38
rs1032336512 Chr.9: 136845674-136845674 on Build GRCh38
rs765793499 Chr.9: 136845775-136845775 on Build GRCh38
rs770693489 Chr.9: 136845712-136845712 on Build GRCh38
rs778221852 Chr.9: 136845690-136845690 on Build GRCh38
rs764134799 Chr.9: 136845619-136845619 on Build GRCh38
rs776614631 Chr.9: 136845716-136845716 on Build GRCh38
rs1018741444 Chr.9: 136845727-136845727 on Build GRCh38
rs756515568 Chr.9: 136845662-136845662 on Build GRCh38
rs763080457 Chr.9: 136845610-136845610 on Build GRCh38
rs751670859 Chr.9: 136845629-136845629 on Build GRCh38
rs755524596 Chr.9: 136845683-136845683 on Build GRCh38
rs780607139 Chr.9: 136845664-136845664 on Build GRCh38
rs999467034 Chr.9: 136845673-136845673 on Build GRCh38
rs777563591 Chr.9: 136845784-136845784 on Build GRCh38
rs767698977 Chr.9: 136845735-136845735 on Build GRCh38
rs755402093 Chr.9: 136845774-136845774 on Build GRCh38
rs745731145 Chr.9: 136845719-136845719 on Build GRCh38
rs977302415 Chr.9: 136845754-136845754 on Build GRCh38
rs767752100 Chr.9: 136845638-136845638 on Build GRCh38
rs762091883 Chr.9: 136845700-136845701 on Build GRCh38
rs184306980 Chr.9: 136845671-136845671 on Build GRCh38
rs758021856 Chr.9: 136845781-136845781 on Build GRCh38
rs761986244 Chr.9: 136845632-136845632 on Build GRCh38

View Full Product Details