Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138140027-138140298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD8
Entrez Gene ID
10902
Gene Name
bromodomain containing 8
Gene Aliases
SMAP, SMAP2, p120
UniGene
Hs.519337
Gene Chromosome Location
Chr.5: 138139766-138178669 on Build GRCh38
Transcripts
NM_139199.1 XM_005271855.2 XM_005271856.2 XM_005271857.2 XM_005271859.3 XM_011543109.1 XM_011543110.1 XM_017008969.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3850365 c.3616G>A p.V1206I Chr.5: 138140166-138140166 on Build GRCh38
5996162 c.3701T>C p.V1234A Chr.5: 138140081-138140081 on Build GRCh38
1696218 c.3679_3680CC>TT p.P1227L Chr.5: 138140102-138140103 on Build GRCh38
5720372 c.3665G>A p.G1222E Chr.5: 138140117-138140117 on Build GRCh38
3428934 c.3641G>T p.R1214I Chr.5: 138140141-138140141 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971468241 Chr.5: 138140069-138140069 on Build GRCh38
rs116327784 Chr.5: 138140257-138140257 on Build GRCh38
rs759769084 Chr.5: 138140140-138140140 on Build GRCh38
rs576422409 Chr.5: 138140260-138140260 on Build GRCh38
rs755620694 Chr.5: 138140100-138140100 on Build GRCh38
rs751381811 Chr.5: 138140204-138140204 on Build GRCh38
rs933513460 Chr.5: 138140270-138140270 on Build GRCh38
rs776950372 Chr.5: 138140136-138140136 on Build GRCh38
rs914795171 Chr.5: 138140162-138140162 on Build GRCh38
rs753490135 Chr.5: 138140196-138140196 on Build GRCh38
rs423258 Chr.5: 138140098-138140098 on Build GRCh38
rs765279438 Chr.5: 138140146-138140146 on Build GRCh38
rs747319280 Chr.5: 138140119-138140119 on Build GRCh38
rs756919597 Chr.5: 138140092-138140092 on Build GRCh38
rs775623472 Chr.5: 138140161-138140161 on Build GRCh38
rs763541719 Chr.5: 138140195-138140195 on Build GRCh38
rs762806835 Chr.5: 138140173-138140173 on Build GRCh38
rs757380718 Chr.5: 138140207-138140213 on Build GRCh38
rs751291624 Chr.5: 138140081-138140081 on Build GRCh38
rs199522004 Chr.5: 138140111-138140111 on Build GRCh38
rs760161673 Chr.5: 138140112-138140112 on Build GRCh38
rs764037114 Chr.5: 138140186-138140186 on Build GRCh38
rs571968530 Chr.5: 138140130-138140130 on Build GRCh38
rs757009806 Chr.5: 138140212-138140212 on Build GRCh38
rs535875927 Chr.5: 138140239-138140239 on Build GRCh38
rs771299574 Chr.5: 138140122-138140122 on Build GRCh38
rs767269726 Chr.5: 138140215-138140215 on Build GRCh38
rs773714833 Chr.5: 138140118-138140118 on Build GRCh38
rs763850003 Chr.5: 138140064-138140064 on Build GRCh38

View Full Product Details