Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 95438966-95439237 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNE3
Entrez Gene ID
161176
Gene Name
spectrin repeat containing nuclear envelope family member 3
Gene Aliases
C14orf49, NET53, Nesp3
UniGene
Hs.354645
Gene Chromosome Location
Chr.14: 95416083-95516693 on Build GRCh38
Transcripts
NM_152592.3 XM_005267377.3 XM_006720063.3 XM_011536515.2 XM_017021056.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1147430 c.2374C>T p.R792C Chr.14: 95439035-95439035 on Build GRCh38
7133162 c.2375G>A p.R792H Chr.14: 95439034-95439034 on Build GRCh38
699729 c.2374C>T p.R792C Chr.14: 95439035-95439035 on Build GRCh38
6438485 c.2344_2345CC>TT p.P782F Chr.14: 95439064-95439065 on Build GRCh38
2253857 c.2376+1G>C p.? Chr.14: 95439032-95439032 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888791044 Chr.14: 95439211-95439211 on Build GRCh38
rs760638317 Chr.14: 95439061-95439061 on Build GRCh38
rs748816553 Chr.14: 95439207-95439207 on Build GRCh38
rs994613784 Chr.14: 95439180-95439180 on Build GRCh38
rs550965393 Chr.14: 95439082-95439082 on Build GRCh38
rs762195083 Chr.14: 95438990-95438990 on Build GRCh38
rs755767118 Chr.14: 95439200-95439200 on Build GRCh38
rs774870853 Chr.14: 95439096-95439096 on Build GRCh38
rs372466452 Chr.14: 95438987-95438987 on Build GRCh38
rs773702748 Chr.14: 95439108-95439108 on Build GRCh38
rs575658188 Chr.14: 95439067-95439067 on Build GRCh38
rs113759233 Chr.14: 95439083-95439083 on Build GRCh38
rs759033948 Chr.14: 95439149-95439149 on Build GRCh38
rs548538141 Chr.14: 95439189-95439196 on Build GRCh38
rs530106374 Chr.14: 95439203-95439203 on Build GRCh38
rs146407791 Chr.14: 95439040-95439040 on Build GRCh38
rs750703159 Chr.14: 95438993-95438993 on Build GRCh38
rs373563721 Chr.14: 95438999-95438999 on Build GRCh38
rs897958721 Chr.14: 95439135-95439135 on Build GRCh38
rs201711548 Chr.14: 95439054-95439054 on Build GRCh38
rs751557832 Chr.14: 95439148-95439148 on Build GRCh38
rs750161867 Chr.14: 95439172-95439172 on Build GRCh38
rs781099784 Chr.14: 95439150-95439150 on Build GRCh38
rs753950082 Chr.14: 95439024-95439024 on Build GRCh38
rs779552067 Chr.14: 95439060-95439060 on Build GRCh38
rs777728702 Chr.14: 95439035-95439035 on Build GRCh38
rs773168780 Chr.14: 95438989-95438989 on Build GRCh38
rs377209700 Chr.14: 95439076-95439076 on Build GRCh38
rs555821802 Chr.14: 95439064-95439064 on Build GRCh38
rs925944304 Chr.14: 95439032-95439032 on Build GRCh38
rs765212058 Chr.14: 95439121-95439121 on Build GRCh38
rs141609511 Chr.14: 95439097-95439097 on Build GRCh38
rs149622983 Chr.14: 95439120-95439120 on Build GRCh38
rs769423169 Chr.14: 95439056-95439056 on Build GRCh38
rs978203906 Chr.14: 95439094-95439094 on Build GRCh38
rs767653566 Chr.14: 95438992-95438992 on Build GRCh38
rs770818140 Chr.14: 95439074-95439074 on Build GRCh38
rs554951423 Chr.14: 95439041-95439041 on Build GRCh38
rs772524614 Chr.14: 95439102-95439102 on Build GRCh38
rs757125407 Chr.14: 95439038-95439038 on Build GRCh38
rs766645599 Chr.14: 95439118-95439118 on Build GRCh38
rs144319184 Chr.14: 95439147-95439147 on Build GRCh38
rs751335846 Chr.14: 95439037-95439037 on Build GRCh38
rs761031540 Chr.14: 95439114-95439114 on Build GRCh38
rs974653250 Chr.14: 95439009-95439009 on Build GRCh38
rs758419026 Chr.14: 95439034-95439034 on Build GRCh38

View Full Product Details