Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3755416-3755689 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf27
Entrez Gene ID
54976
Gene Name
chromosome 20 open reading frame 27
Gene Aliases
-
UniGene
Hs.274422
Gene Chromosome Location
Chr.20: 3753499-3768388 on Build GRCh38
Transcripts
NM_001039140.2 NM_001258429.1 NM_001258430.1 NR_047675.1 XM_011529266.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4098294 c.316C>T p.R106C Chr.20: 3755529-3755529 on Build GRCh38
4901362 c.330C>T p.V110V Chr.20: 3755515-3755515 on Build GRCh38
5981653 c.317G>A p.R106H Chr.20: 3755528-3755528 on Build GRCh38
4581795 c.296A>C p.H99P Chr.20: 3755549-3755549 on Build GRCh38
7149081 c.270G>T p.E90D Chr.20: 3755575-3755575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019160431 Chr.20: 3755508-3755508 on Build GRCh38
rs772082437 Chr.20: 3755436-3755436 on Build GRCh38
rs773439619 Chr.20: 3755659-3755659 on Build GRCh38
rs768818146 Chr.20: 3755534-3755534 on Build GRCh38
rs758412941 Chr.20: 3755525-3755525 on Build GRCh38
rs940306865 Chr.20: 3755459-3755459 on Build GRCh38
rs762101452 Chr.20: 3755542-3755542 on Build GRCh38
rs753563890 Chr.20: 3755503-3755503 on Build GRCh38
rs543589017 Chr.20: 3755527-3755527 on Build GRCh38
rs149371540 Chr.20: 3755529-3755529 on Build GRCh38
rs138776644 Chr.20: 3755526-3755526 on Build GRCh38
rs774731202 Chr.20: 3755536-3755536 on Build GRCh38
rs761086168 Chr.20: 3755569-3755569 on Build GRCh38
rs201614640 Chr.20: 3755626-3755626 on Build GRCh38
rs764914283 Chr.20: 3755581-3755581 on Build GRCh38
rs762332263 Chr.20: 3755479-3755479 on Build GRCh38
rs554641319 Chr.20: 3755488-3755488 on Build GRCh38
rs778870745 Chr.20: 3755505-3755505 on Build GRCh38
rs749409259 Chr.20: 3755612-3755613 on Build GRCh38
rs745997927 Chr.20: 3755578-3755578 on Build GRCh38
rs754651420 Chr.20: 3755504-3755504 on Build GRCh38
rs752552293 Chr.20: 3755599-3755599 on Build GRCh38
rs773830559 Chr.20: 3755563-3755563 on Build GRCh38
rs150224126 Chr.20: 3755518-3755518 on Build GRCh38
rs779053037 Chr.20: 3755656-3755656 on Build GRCh38
rs765797862 Chr.20: 3755500-3755500 on Build GRCh38
rs774677917 Chr.20: 3755478-3755478 on Build GRCh38
rs922195967 Chr.20: 3755662-3755662 on Build GRCh38
rs753235419 Chr.20: 3755575-3755575 on Build GRCh38
rs772522723 Chr.20: 3755658-3755658 on Build GRCh38
rs369413607 Chr.20: 3755440-3755440 on Build GRCh38
rs749328352 Chr.20: 3755634-3755634 on Build GRCh38
rs765959269 Chr.20: 3755572-3755572 on Build GRCh38
rs757119841 Chr.20: 3755622-3755622 on Build GRCh38
rs200138100 Chr.20: 3755624-3755624 on Build GRCh38
rs148088379 Chr.20: 3755487-3755487 on Build GRCh38
rs140915844 Chr.20: 3755513-3755513 on Build GRCh38
rs373108034 Chr.20: 3755616-3755616 on Build GRCh38
rs542910804 Chr.20: 3755439-3755439 on Build GRCh38
rs775839896 Chr.20: 3755456-3755456 on Build GRCh38
rs500247 Chr.20: 3755657-3755657 on Build GRCh38
rs764482812 Chr.20: 3755465-3755465 on Build GRCh38
rs775752981 Chr.20: 3755609-3755610 on Build GRCh38
rs372343006 Chr.20: 3755528-3755528 on Build GRCh38
rs548455526 Chr.20: 3755635-3755635 on Build GRCh38
rs776603873 Chr.20: 3755438-3755438 on Build GRCh38
rs763157999 Chr.20: 3755464-3755464 on Build GRCh38
rs185503510 Chr.20: 3755549-3755549 on Build GRCh38
rs141851694 Chr.20: 3755617-3755617 on Build GRCh38
rs767973515 Chr.20: 3755484-3755484 on Build GRCh38

View Full Product Details