Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 96013713-96013982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CC2D2B
Entrez Gene ID
387707
Gene Name
coiled-coil and C2 domain containing 2B
Gene Aliases
C10orf130, bA248J23.4
UniGene
Hs.538374
Gene Chromosome Location
Chr.10: 95974029-96035167 on Build GRCh38
Transcripts
NM_001001732.3 NM_001159747.1 XM_011539789.2 XM_011539790.2 XM_011539791.2 XM_011539792.2 XM_011539793.2 XM_011539795.2 XM_017016240.1 NR_038444.1

Gene Symbol
ENTPD1-AS1
Entrez Gene ID
728558
Gene Name
ENTPD1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.538374
Gene Chromosome Location
Chr.10: 95753206-96090235 on Build GRCh38
Transcripts
NM_001001732.3 NM_001159747.1 XM_011539789.2 XM_011539790.2 XM_011539791.2 XM_011539792.2 XM_011539793.2 XM_011539795.2 XM_017016240.1 NR_038444.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
921847 c.322C>A p.L108I Chr.10: 96013791-96013791 on Build GRCh38
1585172 c.322C>A p.L108I Chr.10: 96013791-96013791 on Build GRCh38
1603902 c.367C>T p.P123S Chr.10: 96013836-96013836 on Build GRCh38
5341341 c.319-7A>T p.? Chr.10: 96013781-96013781 on Build GRCh38
1603901 c.367C>T p.P123S Chr.10: 96013836-96013836 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778218256 Chr.10: 96013832-96013832 on Build GRCh38
rs772147924 Chr.10: 96013861-96013861 on Build GRCh38
rs974774889 Chr.10: 96013904-96013904 on Build GRCh38
rs756985611 Chr.10: 96013784-96013784 on Build GRCh38
rs760684731 Chr.10: 96013873-96013873 on Build GRCh38
rs759468237 Chr.10: 96013753-96013753 on Build GRCh38
rs537448249 Chr.10: 96013746-96013746 on Build GRCh38
rs959288343 Chr.10: 96013870-96013870 on Build GRCh38
rs558092135 Chr.10: 96013791-96013791 on Build GRCh38
rs368704298 Chr.10: 96013859-96013859 on Build GRCh38
rs1042176795 Chr.10: 96013941-96013941 on Build GRCh38
rs371948458 Chr.10: 96013774-96013774 on Build GRCh38
rs773950220 Chr.10: 96013891-96013892 on Build GRCh38
rs757361702 Chr.10: 96013809-96013809 on Build GRCh38
rs755526424 Chr.10: 96013792-96013792 on Build GRCh38
rs542379350 Chr.10: 96013895-96013895 on Build GRCh38
rs774460048 Chr.10: 96013752-96013752 on Build GRCh38
rs753384479 Chr.10: 96013777-96013777 on Build GRCh38
rs202065763 Chr.10: 96013834-96013834 on Build GRCh38
rs757915544 Chr.10: 96013922-96013922 on Build GRCh38
rs573533124 Chr.10: 96013893-96013893 on Build GRCh38
rs375316468 Chr.10: 96013856-96013856 on Build GRCh38
rs760398440 Chr.10: 96013772-96013772 on Build GRCh38
rs996149014 Chr.10: 96013804-96013804 on Build GRCh38
rs922968267 Chr.10: 96013918-96013918 on Build GRCh38
rs752648077 Chr.10: 96013767-96013767 on Build GRCh38
rs778256705 Chr.10: 96013786-96013786 on Build GRCh38
rs560165236 Chr.10: 96013865-96013865 on Build GRCh38
rs763775803 Chr.10: 96013877-96013877 on Build GRCh38
rs767502871 Chr.10: 96013757-96013757 on Build GRCh38
rs776530961 Chr.10: 96013892-96013892 on Build GRCh38
rs748668339 Chr.10: 96013801-96013801 on Build GRCh38
rs146912146 Chr.10: 96013797-96013797 on Build GRCh38
rs984283532 Chr.10: 96013808-96013808 on Build GRCh38

View Full Product Details