Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 74589510-74589771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERICH3
Entrez Gene ID
127254
Gene Name
glutamate rich 3
Gene Aliases
C1orf173
UniGene
Hs.531182
Gene Chromosome Location
Chr.1: 74568111-74674343 on Build GRCh38
Transcripts
NM_001002912.4 XM_017000274.1 XM_017000275.1 NR_121670.1 NR_121671.1

Gene Symbol
ERICH3-AS1
Entrez Gene ID
101927320
Gene Name
ERICH3 antisense RNA 1
Gene Aliases
-
UniGene
Hs.662589
Gene Chromosome Location
Chr.1: 74577430-74626098 on Build GRCh38
Transcripts
NM_001002912.4 XM_017000274.1 XM_017000275.1 NR_121670.1 NR_121671.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5684388 c.1525delG p.E509fs*7 Chr.1: 74589691-74589691 on Build GRCh38
7251501 c.1569T>A p.P523P Chr.1: 74589647-74589647 on Build GRCh38
4477495 c.2159C>T p.P720L Chr.1: 74589648-74589648 on Build GRCh38
4892195 c.1465G>A p.E489K Chr.1: 74589751-74589751 on Build GRCh38
5684387 c.2116delG p.E706fs*7 Chr.1: 74589691-74589691 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751045981 Chr.1: 74589734-74589734 on Build GRCh38
rs973260322 Chr.1: 74589602-74589602 on Build GRCh38
rs780916856 Chr.1: 74589737-74589737 on Build GRCh38
rs766339093 Chr.1: 74589573-74589573 on Build GRCh38
rs527911768 Chr.1: 74589649-74589649 on Build GRCh38
rs926288824 Chr.1: 74589562-74589562 on Build GRCh38
rs189774313 Chr.1: 74589597-74589597 on Build GRCh38
rs559905474 Chr.1: 74589715-74589715 on Build GRCh38
rs139039758 Chr.1: 74589640-74589640 on Build GRCh38
rs539124280 Chr.1: 74589616-74589616 on Build GRCh38
rs147014389 Chr.1: 74589679-74589679 on Build GRCh38
rs778914098 Chr.1: 74589609-74589609 on Build GRCh38
rs562841225 Chr.1: 74589565-74589565 on Build GRCh38
rs759455508 Chr.1: 74589702-74589702 on Build GRCh38
rs143871156 Chr.1: 74589741-74589741 on Build GRCh38
rs1033423593 Chr.1: 74589570-74589570 on Build GRCh38
rs770254040 Chr.1: 74589742-74589742 on Build GRCh38
rs200867600 Chr.1: 74589619-74589619 on Build GRCh38
rs201084674 Chr.1: 74589629-74589629 on Build GRCh38
rs570011589 Chr.1: 74589591-74589591 on Build GRCh38
rs1054543 Chr.1: 74589618-74589618 on Build GRCh38
rs187355278 Chr.1: 74589743-74589743 on Build GRCh38
rs560448422 Chr.1: 74589648-74589648 on Build GRCh38
rs758026394 Chr.1: 74589600-74589600 on Build GRCh38
rs546358770 Chr.1: 74589715-74589715 on Build GRCh38
rs765046970 Chr.1: 74589708-74589708 on Build GRCh38
rs1049376580 Chr.1: 74589644-74589644 on Build GRCh38
rs977264634 Chr.1: 74589665-74589665 on Build GRCh38
rs772215438 Chr.1: 74589645-74589645 on Build GRCh38
rs950613726 Chr.1: 74589612-74589612 on Build GRCh38
rs753566692 Chr.1: 74589574-74589574 on Build GRCh38
rs775759014 Chr.1: 74589646-74589646 on Build GRCh38
rs138086717 Chr.1: 74589698-74589698 on Build GRCh38
rs752387660 Chr.1: 74589598-74589598 on Build GRCh38
rs888499709 Chr.1: 74589647-74589647 on Build GRCh38
rs921354751 Chr.1: 74589723-74589723 on Build GRCh38
rs746436064 Chr.1: 74589630-74589630 on Build GRCh38
rs267598718 Chr.1: 74589681-74589681 on Build GRCh38
rs867387777 Chr.1: 74589709-74589709 on Build GRCh38
rs761890198 Chr.1: 74589682-74589682 on Build GRCh38
rs758018838 Chr.1: 74589711-74589711 on Build GRCh38
rs980438385 Chr.1: 74589540-74589540 on Build GRCh38
rs906186183 Chr.1: 74589556-74589556 on Build GRCh38
rs944005127 Chr.1: 74589638-74589638 on Build GRCh38
rs2305549 Chr.1: 74589735-74589735 on Build GRCh38
rs766243088 Chr.1: 74589691-74589691 on Build GRCh38
rs777732334 Chr.1: 74589710-74589711 on Build GRCh38
rs752483212 Chr.1: 74589710-74589710 on Build GRCh38

View Full Product Details