Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 108136982-108137255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG4A
Entrez Gene ID
115201
Gene Name
autophagy related 4A cysteine peptidase
Gene Aliases
APG4A, AUTL2
UniGene
Hs.8763
Gene Chromosome Location
Chr.X: 108088900-108154671 on Build GRCh38
Transcripts
NM_001321287.1 NM_001321288.1 NM_001321289.1 NM_001321290.1 NM_052936.4 NM_178270.3 NM_178271.2 NR_135608.1 XM_011530841.2 XM_011530842.1

SNP Detail

SNP ID Chromosome Location
rs763200791 Chr.X: 108137120-108137120 on Build GRCh38
rs79788197 Chr.X: 108137163-108137163 on Build GRCh38
rs146980867 Chr.X: 108137134-108137134 on Build GRCh38
rs940403789 Chr.X: 108137011-108137011 on Build GRCh38
rs1036436059 Chr.X: 108137014-108137014 on Build GRCh38
rs369504594 Chr.X: 108137189-108137189 on Build GRCh38
rs765354592 Chr.X: 108137075-108137075 on Build GRCh38
rs771208478 Chr.X: 108137201-108137201 on Build GRCh38
rs766273007 Chr.X: 108137133-108137133 on Build GRCh38
rs745694868 Chr.X: 108137069-108137069 on Build GRCh38
rs747774203 Chr.X: 108137150-108137150 on Build GRCh38
rs775498678 Chr.X: 108137177-108137178 on Build GRCh38
rs752880103 Chr.X: 108137216-108137216 on Build GRCh38
rs898794102 Chr.X: 108137049-108137049 on Build GRCh38
rs778617203 Chr.X: 108137139-108137139 on Build GRCh38
rs750183063 Chr.X: 108137128-108137128 on Build GRCh38
rs781712105 Chr.X: 108137213-108137213 on Build GRCh38
rs147791568 Chr.X: 108137113-108137113 on Build GRCh38
rs754499020 Chr.X: 108137136-108137136 on Build GRCh38
rs1043458708 Chr.X: 108137145-108137145 on Build GRCh38
rs889308082 Chr.X: 108137179-108137179 on Build GRCh38
rs769557336 Chr.X: 108137084-108137084 on Build GRCh38
rs141263188 Chr.X: 108137124-108137124 on Build GRCh38
rs755874196 Chr.X: 108137132-108137132 on Build GRCh38
rs536490506 Chr.X: 108137050-108137050 on Build GRCh38
rs142072247 Chr.X: 108137109-108137109 on Build GRCh38
rs759535329 Chr.X: 108137009-108137009 on Build GRCh38
rs759653096 Chr.X: 108137056-108137056 on Build GRCh38
rs747344599 Chr.X: 108137198-108137198 on Build GRCh38

View Full Product Details