Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 177513676-177513905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX41
Entrez Gene ID
51428
Gene Name
DEAD-box helicase 41
Gene Aliases
ABS, MPLPF
UniGene
Hs.484288
Gene Chromosome Location
Chr.5: 177511577-177517326 on Build GRCh38
Transcripts
NM_001321732.1 NM_001321830.1 NM_016222.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1436357 c.1063G>A p.E355K Chr.5: 177513720-177513720 on Build GRCh38
4935220 c.1075C>A p.R359S Chr.5: 177513708-177513708 on Build GRCh38
6641819 c.1072A>G p.I358V Chr.5: 177513711-177513711 on Build GRCh38
5817047 c.984G>A p.L328L Chr.5: 177513799-177513799 on Build GRCh38
310513 c.987G>C p.Q329H Chr.5: 177513796-177513796 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370129260 Chr.5: 177513750-177513750 on Build GRCh38
rs199581668 Chr.5: 177513721-177513721 on Build GRCh38
rs879118542 Chr.5: 177513872-177513872 on Build GRCh38
rs775995558 Chr.5: 177513856-177513856 on Build GRCh38
rs772903008 Chr.5: 177513848-177513848 on Build GRCh38
rs77955864 Chr.5: 177513876-177513876 on Build GRCh38
rs749551986 Chr.5: 177513706-177513706 on Build GRCh38
rs776202499 Chr.5: 177513754-177513754 on Build GRCh38
rs538381233 Chr.5: 177513751-177513751 on Build GRCh38
rs556677138 Chr.5: 177513757-177513757 on Build GRCh38
rs764270601 Chr.5: 177513773-177513773 on Build GRCh38
rs546179208 Chr.5: 177513874-177513874 on Build GRCh38
rs754052450 Chr.5: 177513799-177513799 on Build GRCh38
rs758175653 Chr.5: 177513831-177513831 on Build GRCh38
rs772038708 Chr.5: 177513733-177513733 on Build GRCh38
rs765532349 Chr.5: 177513878-177513878 on Build GRCh38
rs774698335 Chr.5: 177513767-177513767 on Build GRCh38
rs141520272 Chr.5: 177513838-177513838 on Build GRCh38
rs768156226 Chr.5: 177513845-177513845 on Build GRCh38
rs759862062 Chr.5: 177513768-177513768 on Build GRCh38
rs542411494 Chr.5: 177513817-177513817 on Build GRCh38
rs779344903 Chr.5: 177513818-177513818 on Build GRCh38
rs148384288 Chr.5: 177513770-177513770 on Build GRCh38
rs201829616 Chr.5: 177513769-177513769 on Build GRCh38
rs377107791 Chr.5: 177513847-177513847 on Build GRCh38
rs757045460 Chr.5: 177513715-177513715 on Build GRCh38
rs775327364 Chr.5: 177513737-177513737 on Build GRCh38
rs146125027 Chr.5: 177513854-177513854 on Build GRCh38
rs558863910 Chr.5: 177513697-177513697 on Build GRCh38
rs780003390 Chr.5: 177513837-177513837 on Build GRCh38
rs780667117 Chr.5: 177513846-177513846 on Build GRCh38
rs335439 Chr.5: 177513870-177513870 on Build GRCh38
rs267600572 Chr.5: 177513695-177513695 on Build GRCh38
rs778864620 Chr.5: 177513720-177513720 on Build GRCh38
rs752201749 Chr.5: 177513789-177513791 on Build GRCh38
rs200664784 Chr.5: 177513820-177513820 on Build GRCh38

View Full Product Details