Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4409221-4409480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHAF1A
Entrez Gene ID
10036
Gene Name
chromatin assembly factor 1 subunit A
Gene Aliases
CAF-1, CAF1, CAF1B, CAF1P150, P150
UniGene
Hs.79018
Gene Chromosome Location
Chr.19: 4402596-4448322 on Build GRCh38
Transcripts
NM_005483.2 XM_011527605.2 XM_011527606.2 XM_011527607.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4078965 c.554A>C p.E185A Chr.19: 4409353-4409353 on Build GRCh38
4404319 c.593A>C p.D198A Chr.19: 4409392-4409392 on Build GRCh38
997643 c.509C>T p.A170V Chr.19: 4409308-4409308 on Build GRCh38
3535286 c.488C>T p.A163V Chr.19: 4409287-4409287 on Build GRCh38
4607711 c.650G>T p.C217F Chr.19: 4409449-4409449 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764331314 Chr.19: 4409407-4409407 on Build GRCh38
rs758235903 Chr.19: 4409410-4409410 on Build GRCh38
rs754501034 Chr.19: 4409422-4409422 on Build GRCh38
rs138213365 Chr.19: 4409296-4409296 on Build GRCh38
rs776192737 Chr.19: 4409276-4409276 on Build GRCh38
rs766944883 Chr.19: 4409382-4409382 on Build GRCh38
rs779932686 Chr.19: 4409412-4409412 on Build GRCh38
rs745666920 Chr.19: 4409324-4409324 on Build GRCh38
rs776340717 Chr.19: 4409353-4409353 on Build GRCh38
rs201627290 Chr.19: 4409441-4409441 on Build GRCh38
rs747701573 Chr.19: 4409425-4409425 on Build GRCh38
rs758291420 Chr.19: 4409340-4409340 on Build GRCh38
rs752904482 Chr.19: 4409305-4409305 on Build GRCh38
rs752833373 Chr.19: 4409402-4409402 on Build GRCh38
rs750956724 Chr.19: 4409287-4409287 on Build GRCh38
rs750683545 Chr.19: 4409285-4409285 on Build GRCh38
rs767561557 Chr.19: 4409399-4409399 on Build GRCh38
rs746983454 Chr.19: 4409413-4409413 on Build GRCh38
rs376205419 Chr.19: 4409391-4409391 on Build GRCh38
rs199665572 Chr.19: 4409403-4409403 on Build GRCh38
rs183942259 Chr.19: 4409348-4409348 on Build GRCh38
rs200445124 Chr.19: 4409373-4409373 on Build GRCh38
rs950782305 Chr.19: 4409452-4409452 on Build GRCh38
rs554787620 Chr.19: 4409408-4409408 on Build GRCh38
rs763499968 Chr.19: 4409288-4409288 on Build GRCh38
rs773021521 Chr.19: 4409432-4409432 on Build GRCh38
rs1026526574 Chr.19: 4409446-4409446 on Build GRCh38
rs767002305 Chr.19: 4409289-4409289 on Build GRCh38
rs759901021 Chr.19: 4409453-4409453 on Build GRCh38
rs756495652 Chr.19: 4409348-4409350 on Build GRCh38
rs1022490964 Chr.19: 4409251-4409251 on Build GRCh38
rs778512366 Chr.19: 4409322-4409322 on Build GRCh38
rs749649310 Chr.19: 4409440-4409440 on Build GRCh38
rs763446721 Chr.19: 4409376-4409376 on Build GRCh38
rs768089075 Chr.19: 4409351-4409351 on Build GRCh38
rs545280672 Chr.19: 4409309-4409309 on Build GRCh38
rs546330233 Chr.19: 4409297-4409297 on Build GRCh38
rs529839301 Chr.19: 4409286-4409286 on Build GRCh38
rs760205920 Chr.19: 4409260-4409260 on Build GRCh38
rs769503119 Chr.19: 4409374-4409374 on Build GRCh38
rs769295287 Chr.19: 4409431-4409431 on Build GRCh38
rs538212376 Chr.19: 4409341-4409341 on Build GRCh38
rs371637761 Chr.19: 4409359-4409359 on Build GRCh38
rs768303127 Chr.19: 4409274-4409274 on Build GRCh38
rs780512913 Chr.19: 4409423-4409423 on Build GRCh38
rs768072344 Chr.19: 4409456-4409456 on Build GRCh38
rs188842259 Chr.19: 4409390-4409390 on Build GRCh38
rs35651457 Chr.19: 4409299-4409299 on Build GRCh38
rs774996487 Chr.19: 4409448-4409448 on Build GRCh38
rs373856871 Chr.19: 4409249-4409249 on Build GRCh38

View Full Product Details