Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 83963144-83963405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDIL3
Entrez Gene ID
10085
Gene Name
EGF like repeats and discoidin domains 3
Gene Aliases
DEL1
UniGene
Hs.482730
Gene Chromosome Location
Chr.5: 83940595-84384867 on Build GRCh38
Transcripts
NM_001278642.1 NM_005711.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5356610 c.1288G>T p.D430Y Chr.5: 83963210-83963210 on Build GRCh38
4806228 c.1256G>T p.W419L Chr.5: 83963242-83963242 on Build GRCh38
7210731 c.1170C>T p.G390G Chr.5: 83963328-83963328 on Build GRCh38
1642803 c.1138-4delT p.? Chr.5: 83963364-83963364 on Build GRCh38
5868910 c.1184G>A p.G395E Chr.5: 83963314-83963314 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772050783 Chr.5: 83963285-83963285 on Build GRCh38
rs368407385 Chr.5: 83963336-83963336 on Build GRCh38
rs536188516 Chr.5: 83963262-83963262 on Build GRCh38
rs753299184 Chr.5: 83963363-83963364 on Build GRCh38
rs768315119 Chr.5: 83963274-83963274 on Build GRCh38
rs750778651 Chr.5: 83963372-83963372 on Build GRCh38
rs188543397 Chr.5: 83963364-83963364 on Build GRCh38
rs773145193 Chr.5: 83963363-83963363 on Build GRCh38
rs374240916 Chr.5: 83963246-83963246 on Build GRCh38
rs973864529 Chr.5: 83963294-83963294 on Build GRCh38
rs552874498 Chr.5: 83963339-83963339 on Build GRCh38
rs757509169 Chr.5: 83963220-83963220 on Build GRCh38
rs906772255 Chr.5: 83963359-83963359 on Build GRCh38
rs28590009 Chr.5: 83963354-83963354 on Build GRCh38
rs776578953 Chr.5: 83963340-83963340 on Build GRCh38
rs764175849 Chr.5: 83963351-83963351 on Build GRCh38
rs778649899 Chr.5: 83963275-83963275 on Build GRCh38
rs910658098 Chr.5: 83963244-83963244 on Build GRCh38
rs368525093 Chr.5: 83963188-83963188 on Build GRCh38
rs767694106 Chr.5: 83963371-83963371 on Build GRCh38
rs563082099 Chr.5: 83963370-83963370 on Build GRCh38
rs773045150 Chr.5: 83963319-83963319 on Build GRCh38
rs267600721 Chr.5: 83963314-83963314 on Build GRCh38
rs753840831 Chr.5: 83963248-83963248 on Build GRCh38
rs919437494 Chr.5: 83963327-83963327 on Build GRCh38
rs541691786 Chr.5: 83963379-83963379 on Build GRCh38
rs781625915 Chr.5: 83963241-83963241 on Build GRCh38
rs747805136 Chr.5: 83963292-83963292 on Build GRCh38
rs140436608 Chr.5: 83963360-83963360 on Build GRCh38
rs748221237 Chr.5: 83963258-83963258 on Build GRCh38
rs779401836 Chr.5: 83963364-83963364 on Build GRCh38
rs751799201 Chr.5: 83963215-83963215 on Build GRCh38
rs558194262 Chr.5: 83963247-83963247 on Build GRCh38
rs758508735 Chr.5: 83963202-83963202 on Build GRCh38
rs1029510228 Chr.5: 83963185-83963185 on Build GRCh38
rs986079792 Chr.5: 83963232-83963232 on Build GRCh38
rs762969320 Chr.5: 83963350-83963350 on Build GRCh38
rs201245990 Chr.5: 83963334-83963334 on Build GRCh38
rs377140824 Chr.5: 83963253-83963253 on Build GRCh38
rs764257952 Chr.5: 83963205-83963205 on Build GRCh38
rs771688663 Chr.5: 83963293-83963293 on Build GRCh38
rs374374243 Chr.5: 83963179-83963179 on Build GRCh38

View Full Product Details