Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 31131441-31131680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSC1
Entrez Gene ID
1823
Gene Name
desmocollin 1
Gene Aliases
CDHF1, DG2/DG3
UniGene
Hs.567260
Gene Chromosome Location
Chr.18: 31129236-31162856 on Build GRCh38
Transcripts
NM_004948.3 NM_024421.2 NR_110785.1

Gene Symbol
DSCAS
Entrez Gene ID
101927698
Gene Name
DSC1/DSC2 antisense RNA
Gene Aliases
-
UniGene
Hs.579538
Gene Chromosome Location
Chr.18: 31101588-31162789 on Build GRCh38
Transcripts
NM_004948.3 NM_024421.2 NR_110785.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5678152 c.2478G>T p.R826R Chr.18: 31131603-31131603 on Build GRCh38
5678151 c.2478G>T p.R826R Chr.18: 31131603-31131603 on Build GRCh38
6083158 c.2412G>A p.K804K Chr.18: 31131669-31131669 on Build GRCh38
6083159 c.2412G>A p.K804K Chr.18: 31131669-31131669 on Build GRCh38
6880794 c.2405C>T p.S802F Chr.18: 31131676-31131676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs16961333 Chr.18: 31131541-31131541 on Build GRCh38
rs750003152 Chr.18: 31131634-31131634 on Build GRCh38
rs373873191 Chr.18: 31131587-31131587 on Build GRCh38
rs764052606 Chr.18: 31131597-31131597 on Build GRCh38
rs16961331 Chr.18: 31131482-31131482 on Build GRCh38
rs780784082 Chr.18: 31131559-31131559 on Build GRCh38
rs749754325 Chr.18: 31131644-31131644 on Build GRCh38
rs764697761 Chr.18: 31131646-31131646 on Build GRCh38
rs779266104 Chr.18: 31131653-31131653 on Build GRCh38
rs755308123 Chr.18: 31131650-31131650 on Build GRCh38
rs769024874 Chr.18: 31131594-31131594 on Build GRCh38
rs774176162 Chr.18: 31131465-31131465 on Build GRCh38
rs370393725 Chr.18: 31131620-31131620 on Build GRCh38
rs267605149 Chr.18: 31131629-31131629 on Build GRCh38
rs996822159 Chr.18: 31131637-31131637 on Build GRCh38
rs146414667 Chr.18: 31131631-31131631 on Build GRCh38
rs377191170 Chr.18: 31131639-31131639 on Build GRCh38
rs758184778 Chr.18: 31131545-31131545 on Build GRCh38
rs886922717 Chr.18: 31131617-31131617 on Build GRCh38
rs765202886 Chr.18: 31131618-31131618 on Build GRCh38
rs987746876 Chr.18: 31131522-31131522 on Build GRCh38
rs745368809 Chr.18: 31131562-31131562 on Build GRCh38
rs1041518862 Chr.18: 31131621-31131621 on Build GRCh38
rs201513884 Chr.18: 31131605-31131605 on Build GRCh38
rs114274263 Chr.18: 31131601-31131601 on Build GRCh38
rs561384554 Chr.18: 31131478-31131478 on Build GRCh38
rs967179464 Chr.18: 31131515-31131515 on Build GRCh38
rs777324060 Chr.18: 31131547-31131547 on Build GRCh38
rs770483568 Chr.18: 31131579-31131579 on Build GRCh38
rs760727264 Chr.18: 31131614-31131614 on Build GRCh38
rs763895775 Chr.18: 31131623-31131623 on Build GRCh38
rs774457500 Chr.18: 31131596-31131596 on Build GRCh38
rs1021528783 Chr.18: 31131512-31131512 on Build GRCh38
rs1789072 Chr.18: 31131556-31131556 on Build GRCh38
rs535797464 Chr.18: 31131584-31131584 on Build GRCh38
rs755652280 Chr.18: 31131636-31131636 on Build GRCh38
rs988340881 Chr.18: 31131504-31131504 on Build GRCh38
rs946084806 Chr.18: 31131544-31131544 on Build GRCh38
rs550227268 Chr.18: 31131550-31131550 on Build GRCh38
rs566691934 Chr.18: 31131658-31131658 on Build GRCh38
rs776154525 Chr.18: 31131468-31131468 on Build GRCh38
rs757001906 Chr.18: 31131630-31131630 on Build GRCh38
rs192144667 Chr.18: 31131506-31131506 on Build GRCh38
rs975112245 Chr.18: 31131483-31131483 on Build GRCh38
rs931187626 Chr.18: 31131604-31131604 on Build GRCh38
rs571791058 Chr.18: 31131523-31131523 on Build GRCh38
rs748455000 Chr.18: 31131655-31131655 on Build GRCh38

View Full Product Details