Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 53821580-53821837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHDH
Entrez Gene ID
55349
Gene Name
choline dehydrogenase
Gene Aliases
-
UniGene
Hs.126688
Gene Chromosome Location
Chr.3: 53813130-53846393 on Build GRCh38
Transcripts
NM_018397.4 XM_006713250.3 XM_006713251.3 XM_006713252.3 XM_011533938.2 XM_011533939.2 XM_017006797.1 XM_017006798.1 XM_017006799.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5059877 c.938C>T p.A313V Chr.3: 53821694-53821694 on Build GRCh38
1047327 c.902C>A p.S301Y Chr.3: 53821730-53821730 on Build GRCh38
6885565 c.890G>A p.G297D Chr.3: 53821742-53821742 on Build GRCh38
6885566 c.889G>A p.G297S Chr.3: 53821743-53821743 on Build GRCh38
5016932 c.946C>G p.L316V Chr.3: 53821686-53821686 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369923460 Chr.3: 53821742-53821742 on Build GRCh38
rs941949328 Chr.3: 53821608-53821608 on Build GRCh38
rs140594688 Chr.3: 53821647-53821647 on Build GRCh38
rs765292151 Chr.3: 53821805-53821805 on Build GRCh38
rs533752759 Chr.3: 53821724-53821724 on Build GRCh38
rs762595515 Chr.3: 53821612-53821612 on Build GRCh38
rs547544165 Chr.3: 53821786-53821786 on Build GRCh38
rs35064726 Chr.3: 53821711-53821711 on Build GRCh38
rs763907297 Chr.3: 53821622-53821622 on Build GRCh38
rs372502015 Chr.3: 53821795-53821795 on Build GRCh38
rs371028326 Chr.3: 53821656-53821656 on Build GRCh38
rs772986165 Chr.3: 53821766-53821766 on Build GRCh38
rs945939054 Chr.3: 53821717-53821717 on Build GRCh38
rs889163968 Chr.3: 53821736-53821736 on Build GRCh38
rs756315133 Chr.3: 53821649-53821649 on Build GRCh38
rs778878023 Chr.3: 53821686-53821686 on Build GRCh38
rs753830426 Chr.3: 53821661-53821661 on Build GRCh38
rs1056830571 Chr.3: 53821701-53821701 on Build GRCh38
rs773766584 Chr.3: 53821737-53821737 on Build GRCh38
rs767498767 Chr.3: 53821740-53821740 on Build GRCh38
rs761679537 Chr.3: 53821739-53821739 on Build GRCh38
rs997497675 Chr.3: 53821702-53821702 on Build GRCh38
rs767261558 Chr.3: 53821645-53821645 on Build GRCh38
rs758960990 Chr.3: 53821809-53821809 on Build GRCh38
rs761680630 Chr.3: 53821639-53821639 on Build GRCh38
rs779131294 Chr.3: 53821612-53821613 on Build GRCh38
rs531526788 Chr.3: 53821631-53821631 on Build GRCh38
rs748646122 Chr.3: 53821687-53821687 on Build GRCh38
rs879889911 Chr.3: 53821611-53821611 on Build GRCh38
rs755185462 Chr.3: 53821799-53821799 on Build GRCh38
rs373320152 Chr.3: 53821725-53821725 on Build GRCh38
rs747439665 Chr.3: 53821715-53821715 on Build GRCh38
rs567672910 Chr.3: 53821807-53821807 on Build GRCh38
rs952129843 Chr.3: 53821609-53821609 on Build GRCh38
rs375478883 Chr.3: 53821727-53821727 on Build GRCh38
rs768164442 Chr.3: 53821735-53821735 on Build GRCh38
rs760502555 Chr.3: 53821772-53821772 on Build GRCh38
rs755131144 Chr.3: 53821673-53821673 on Build GRCh38
rs897807249 Chr.3: 53821684-53821684 on Build GRCh38
rs939376939 Chr.3: 53821696-53821696 on Build GRCh38
rs138124394 Chr.3: 53821660-53821660 on Build GRCh38
rs748726748 Chr.3: 53821733-53821733 on Build GRCh38
rs879021606 Chr.3: 53821689-53821689 on Build GRCh38

View Full Product Details