Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 122834757-122835030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUZD1
Entrez Gene ID
50624
Gene Name
CUB and zona pellucida like domains 1
Gene Aliases
ERG-1, ERG1, ITMAP1, UO-44
UniGene
Hs.647182
Gene Chromosome Location
Chr.10: 122832155-122846175 on Build GRCh38
Transcripts
NM_022034.5 NR_037912.1 NR_037915.1

Gene Symbol
FAM24B-CUZD1
Entrez Gene ID
100533195
Gene Name
FAM24B-CUZD1 readthrough
Gene Aliases
-
UniGene
Hs.647182
Gene Chromosome Location
Chr.10: 122832155-122879641 on Build GRCh38
Transcripts
NM_022034.5 NR_037912.1 NR_037915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2020802 c.1132A>G p.I378V Chr.10: 122834956-122834956 on Build GRCh38
3719096 c.1190T>A p.M397K Chr.10: 122834898-122834898 on Build GRCh38
160288 c.1258C>G p.Q420E Chr.10: 122834830-122834830 on Build GRCh38
4012347 c.1181A>T p.N394I Chr.10: 122834907-122834907 on Build GRCh38
328532 c.1064G>A p.R355H Chr.10: 122835024-122835024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754000450 Chr.10: 122834896-122834896 on Build GRCh38
rs776161317 Chr.10: 122834799-122834799 on Build GRCh38
rs780999461 Chr.10: 122834971-122834971 on Build GRCh38
rs755077881 Chr.10: 122834916-122834916 on Build GRCh38
rs762424447 Chr.10: 122834815-122834815 on Build GRCh38
rs761938782 Chr.10: 122834988-122834988 on Build GRCh38
rs199726363 Chr.10: 122834838-122834838 on Build GRCh38
rs769217392 Chr.10: 122834805-122834805 on Build GRCh38
rs769531909 Chr.10: 122834989-122834989 on Build GRCh38
rs201662828 Chr.10: 122834968-122834968 on Build GRCh38
rs766392941 Chr.10: 122834895-122834895 on Build GRCh38
rs199934875 Chr.10: 122834783-122834783 on Build GRCh38
rs746978782 Chr.10: 122834806-122834806 on Build GRCh38
rs750618478 Chr.10: 122834862-122834862 on Build GRCh38
rs764124363 Chr.10: 122834821-122834834 on Build GRCh38
rs370233601 Chr.10: 122834786-122834786 on Build GRCh38
rs892712444 Chr.10: 122834800-122834800 on Build GRCh38
rs569583802 Chr.10: 122834866-122834866 on Build GRCh38
rs751640179 Chr.10: 122834924-122834925 on Build GRCh38
rs992572658 Chr.10: 122834878-122834878 on Build GRCh38
rs907023707 Chr.10: 122834821-122834821 on Build GRCh38
rs555857606 Chr.10: 122834851-122834851 on Build GRCh38
rs763512650 Chr.10: 122834819-122834819 on Build GRCh38
rs919231799 Chr.10: 122834942-122834942 on Build GRCh38
rs1009932666 Chr.10: 122834802-122834802 on Build GRCh38
rs536247065 Chr.10: 122834841-122834841 on Build GRCh38
rs756322363 Chr.10: 122834875-122834875 on Build GRCh38
rs762227956 Chr.10: 122834849-122834849 on Build GRCh38
rs746885654 Chr.10: 122834950-122834950 on Build GRCh38
rs777925286 Chr.10: 122834946-122834946 on Build GRCh38
rs150494245 Chr.10: 122834801-122834801 on Build GRCh38
rs372927471 Chr.10: 122834782-122834782 on Build GRCh38
rs774616793 Chr.10: 122834940-122834940 on Build GRCh38
rs756933688 Chr.10: 122834909-122834909 on Build GRCh38
rs774971390 Chr.10: 122834991-122834991 on Build GRCh38
rs138479743 Chr.10: 122834960-122834960 on Build GRCh38
rs745526672 Chr.10: 122834986-122834986 on Build GRCh38
rs143015582 Chr.10: 122834970-122834970 on Build GRCh38
rs200686197 Chr.10: 122834839-122834839 on Build GRCh38

View Full Product Details