Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 76439521-76439746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESRRB
Entrez Gene ID
2103
Gene Name
estrogen related receptor beta
Gene Aliases
DFNB35, ERR2, ERRb, ESRL2, NR3B2
UniGene
Hs.435845
Gene Chromosome Location
Chr.14: 76371347-76501839 on Build GRCh38
Transcripts
NM_004452.3 XM_011536547.2 XM_011536550.2 XM_011536553.2 XM_011536554.2 XM_017021085.1 XM_017021086.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5634891 c.340C>A p.H114N Chr.14: 76439693-76439693 on Build GRCh38
3498128 c.180C>T p.P60P Chr.14: 76439533-76439533 on Build GRCh38
6521676 c.197G>A p.G66E Chr.14: 76439550-76439550 on Build GRCh38
6487004 c.319G>A p.G107R Chr.14: 76439672-76439672 on Build GRCh38
958054 c.349G>A p.V117M Chr.14: 76439702-76439702 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376300660 Chr.14: 76439671-76439671 on Build GRCh38
rs559643813 Chr.14: 76439568-76439568 on Build GRCh38
rs755315466 Chr.14: 76439655-76439655 on Build GRCh38
rs756579075 Chr.14: 76439551-76439551 on Build GRCh38
rs770677880 Chr.14: 76439665-76439665 on Build GRCh38
rs780495222 Chr.14: 76439640-76439640 on Build GRCh38
rs769832766 Chr.14: 76439607-76439607 on Build GRCh38
rs567671346 Chr.14: 76439698-76439698 on Build GRCh38
rs121909110 Chr.14: 76439682-76439682 on Build GRCh38
rs771657920 Chr.14: 76439575-76439575 on Build GRCh38
rs761936066 Chr.14: 76439627-76439627 on Build GRCh38
rs757302943 Chr.14: 76439545-76439545 on Build GRCh38
rs750478006 Chr.14: 76439632-76439632 on Build GRCh38
rs549672955 Chr.14: 76439546-76439546 on Build GRCh38
rs139018322 Chr.14: 76439692-76439692 on Build GRCh38
rs373056537 Chr.14: 76439633-76439633 on Build GRCh38
rs767674170 Chr.14: 76439715-76439715 on Build GRCh38
rs571434756 Chr.14: 76439630-76439630 on Build GRCh38
rs780442131 Chr.14: 76439555-76439555 on Build GRCh38
rs528723972 Chr.14: 76439592-76439592 on Build GRCh38
rs118150038 Chr.14: 76439614-76439614 on Build GRCh38
rs754162236 Chr.14: 76439649-76439649 on Build GRCh38
rs376403497 Chr.14: 76439576-76439576 on Build GRCh38
rs150226207 Chr.14: 76439626-76439626 on Build GRCh38
rs141586518 Chr.14: 76439704-76439704 on Build GRCh38
rs370424727 Chr.14: 76439696-76439696 on Build GRCh38
rs779134127 Chr.14: 76439656-76439656 on Build GRCh38
rs190465039 Chr.14: 76439668-76439668 on Build GRCh38
rs771967770 Chr.14: 76439597-76439597 on Build GRCh38
rs775439645 Chr.14: 76439683-76439683 on Build GRCh38
rs775448717 Chr.14: 76439612-76439612 on Build GRCh38
rs1002514503 Chr.14: 76439621-76439621 on Build GRCh38
rs751593912 Chr.14: 76439541-76439541 on Build GRCh38
rs1064438 Chr.14: 76439638-76439638 on Build GRCh38
rs371102435 Chr.14: 76439596-76439596 on Build GRCh38
rs769857569 Chr.14: 76439672-76439672 on Build GRCh38
rs746745430 Chr.14: 76439658-76439658 on Build GRCh38

View Full Product Details