Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53218087-53218360 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf123
Entrez Gene ID
54987
Gene Name
chromosome 1 open reading frame 123
Gene Aliases
-
UniGene
Hs.525391
Gene Chromosome Location
Chr.1: 53214099-53220639 on Build GRCh38
Transcripts
NM_001304759.1 NM_001304760.1 NM_017887.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3419321 c.261+2T>C p.? Chr.1: 53218170-53218170 on Build GRCh38
7076406 c.230-1G>C p.? Chr.1: 53218204-53218204 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs745990853 Chr.1: 53218179-53218179 on Build GRCh38
rs765296408 Chr.1: 53218141-53218141 on Build GRCh38
rs777641694 Chr.1: 53218112-53218112 on Build GRCh38
rs72673130 Chr.1: 53218236-53218236 on Build GRCh38
rs781113815 Chr.1: 53218175-53218175 on Build GRCh38
rs544562776 Chr.1: 53218192-53218192 on Build GRCh38
rs768999922 Chr.1: 53218190-53218190 on Build GRCh38
rs780548676 Chr.1: 53218188-53218188 on Build GRCh38
rs751726732 Chr.1: 53218171-53218171 on Build GRCh38
rs765486105 Chr.1: 53218293-53218293 on Build GRCh38
rs760221907 Chr.1: 53218215-53218215 on Build GRCh38
rs771488417 Chr.1: 53218213-53218214 on Build GRCh38
rs777985329 Chr.1: 53218162-53218162 on Build GRCh38
rs760437711 Chr.1: 53218123-53218123 on Build GRCh38
rs79453977 Chr.1: 53218327-53218327 on Build GRCh38
rs1010450490 Chr.1: 53218265-53218265 on Build GRCh38
rs757561191 Chr.1: 53218173-53218173 on Build GRCh38
rs376808171 Chr.1: 53218122-53218122 on Build GRCh38
rs200312423 Chr.1: 53218229-53218229 on Build GRCh38
rs774833856 Chr.1: 53218197-53218197 on Build GRCh38
rs12140585 Chr.1: 53218249-53218249 on Build GRCh38
rs749762985 Chr.1: 53218206-53218207 on Build GRCh38
rs146156858 Chr.1: 53218305-53218305 on Build GRCh38
rs753436579 Chr.1: 53218136-53218136 on Build GRCh38
rs765789702 Chr.1: 53218125-53218125 on Build GRCh38
rs770249204 Chr.1: 53218186-53218186 on Build GRCh38
rs560506317 Chr.1: 53218285-53218285 on Build GRCh38
rs759513664 Chr.1: 53218137-53218137 on Build GRCh38
rs776731664 Chr.1: 53218180-53218180 on Build GRCh38
rs200743226 Chr.1: 53218145-53218145 on Build GRCh38
rs1039275714 Chr.1: 53218217-53218217 on Build GRCh38
rs763050513 Chr.1: 53218250-53218250 on Build GRCh38
rs866608378 Chr.1: 53218298-53218298 on Build GRCh38
rs752607452 Chr.1: 53218304-53218304 on Build GRCh38
rs999648919 Chr.1: 53218291-53218291 on Build GRCh38
rs775961420 Chr.1: 53218235-53218235 on Build GRCh38
rs758283704 Chr.1: 53218159-53218159 on Build GRCh38
rs902586910 Chr.1: 53218284-53218284 on Build GRCh38

View Full Product Details