Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 36887370-36887629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLDC2
Entrez Gene ID
140711
Gene Name
TBC/LysM-associated domain containing 2
Gene Aliases
C20orf118
UniGene
Hs.472630
Gene Chromosome Location
Chr.20: 36873071-36894235 on Build GRCh38
Transcripts
NM_001304783.1 NM_080628.2 XM_017027672.1 XM_017027673.1 XM_017027674.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4820894 c.464C>G p.S155C Chr.20: 36887480-36887480 on Build GRCh38
6159841 c.497T>C p.M166T Chr.20: 36887513-36887513 on Build GRCh38
5802390 c.463_466delTCTT p.F156fs*2 Chr.20: 36887479-36887482 on Build GRCh38
6802165 c.481G>C p.D161H Chr.20: 36887497-36887497 on Build GRCh38
4984211 c.493C>T p.L165L Chr.20: 36887509-36887509 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764547871 Chr.20: 36887530-36887530 on Build GRCh38
rs544561249 Chr.20: 36887479-36887482 on Build GRCh38
rs559651135 Chr.20: 36887522-36887522 on Build GRCh38
rs372620586 Chr.20: 36887529-36887529 on Build GRCh38
rs763129782 Chr.20: 36887411-36887411 on Build GRCh38
rs757879641 Chr.20: 36887444-36887444 on Build GRCh38
rs757906690 Chr.20: 36887534-36887534 on Build GRCh38
rs371597800 Chr.20: 36887561-36887561 on Build GRCh38
rs543537864 Chr.20: 36887450-36887450 on Build GRCh38
rs770694750 Chr.20: 36887573-36887582 on Build GRCh38
rs754369767 Chr.20: 36887532-36887532 on Build GRCh38
rs754526384 Chr.20: 36887548-36887548 on Build GRCh38
rs747911188 Chr.20: 36887554-36887554 on Build GRCh38
rs150768309 Chr.20: 36887422-36887422 on Build GRCh38
rs756672898 Chr.20: 36887433-36887433 on Build GRCh38
rs780992675 Chr.20: 36887552-36887552 on Build GRCh38
rs753197793 Chr.20: 36887432-36887432 on Build GRCh38
rs374926747 Chr.20: 36887564-36887564 on Build GRCh38
rs773365107 Chr.20: 36887407-36887407 on Build GRCh38
rs747967134 Chr.20: 36887468-36887468 on Build GRCh38
rs1024019382 Chr.20: 36887604-36887604 on Build GRCh38
rs779494085 Chr.20: 36887536-36887536 on Build GRCh38
rs368876056 Chr.20: 36887477-36887477 on Build GRCh38
rs138222765 Chr.20: 36887478-36887478 on Build GRCh38
rs928692721 Chr.20: 36887589-36887589 on Build GRCh38
rs1051184159 Chr.20: 36887439-36887439 on Build GRCh38
rs774602822 Chr.20: 36887505-36887505 on Build GRCh38
rs541072916 Chr.20: 36887488-36887488 on Build GRCh38
rs1014461158 Chr.20: 36887602-36887602 on Build GRCh38
rs746864421 Chr.20: 36887546-36887546 on Build GRCh38
rs369362831 Chr.20: 36887567-36887567 on Build GRCh38
rs769785995 Chr.20: 36887479-36887479 on Build GRCh38
rs940002576 Chr.20: 36887459-36887459 on Build GRCh38
rs755162140 Chr.20: 36887478-36887479 on Build GRCh38
rs754021584 Chr.20: 36887433-36887434 on Build GRCh38
rs759743297 Chr.20: 36887519-36887519 on Build GRCh38
rs376783054 Chr.20: 36887451-36887451 on Build GRCh38
rs368488951 Chr.20: 36887414-36887414 on Build GRCh38
rs778270212 Chr.20: 36887440-36887440 on Build GRCh38
rs755346152 Chr.20: 36887417-36887417 on Build GRCh38
rs779710985 Chr.20: 36887447-36887447 on Build GRCh38
rs766417400 Chr.20: 36887500-36887500 on Build GRCh38
rs746655484 Chr.20: 36887543-36887543 on Build GRCh38
rs756659226 Chr.20: 36887481-36887481 on Build GRCh38
rs776621471 Chr.20: 36887457-36887457 on Build GRCh38
rs750833556 Chr.20: 36887431-36887431 on Build GRCh38
rs767912916 Chr.20: 36887520-36887520 on Build GRCh38

View Full Product Details