Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 39552005-39552278 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM14
Entrez Gene ID
201895
Gene Name
small integral membrane protein 14
Gene Aliases
C4orf34
UniGene
Hs.205952
Gene Chromosome Location
Chr.4: 39547297-39638998 on Build GRCh38
Transcripts
NM_001317896.1 NM_001317897.1 NM_174921.2 XM_011513640.2 XM_017007862.1 XM_017007863.1 NR_047679.1

Gene Symbol
UGDH-AS1
Entrez Gene ID
100885776
Gene Name
UGDH antisense RNA 1
Gene Aliases
-
UniGene
Hs.640769
Gene Chromosome Location
Chr.4: 39527839-39594707 on Build GRCh38
Transcripts
NM_001317896.1 NM_001317897.1 NM_174921.2 XM_011513640.2 XM_017007862.1 XM_017007863.1 NR_047679.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3392859 c.286C>A p.P96T Chr.4: 39552140-39552140 on Build GRCh38
4771660 c.281C>T p.P94L Chr.4: 39552145-39552145 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs961294401 Chr.4: 39552162-39552162 on Build GRCh38
rs752386898 Chr.4: 39552178-39552178 on Build GRCh38
rs910929778 Chr.4: 39552221-39552221 on Build GRCh38
rs753243824 Chr.4: 39552114-39552114 on Build GRCh38
rs773137568 Chr.4: 39552086-39552086 on Build GRCh38
rs1050114951 Chr.4: 39552239-39552239 on Build GRCh38
rs376716371 Chr.4: 39552078-39552078 on Build GRCh38
rs150006812 Chr.4: 39552233-39552233 on Build GRCh38
rs532228895 Chr.4: 39552176-39552176 on Build GRCh38
rs919755584 Chr.4: 39552211-39552211 on Build GRCh38
rs758903063 Chr.4: 39552183-39552183 on Build GRCh38
rs201253169 Chr.4: 39552085-39552085 on Build GRCh38
rs11730245 Chr.4: 39552087-39552087 on Build GRCh38
rs754476811 Chr.4: 39552123-39552123 on Build GRCh38
rs765932779 Chr.4: 39552103-39552103 on Build GRCh38
rs914406965 Chr.4: 39552213-39552213 on Build GRCh38
rs775758097 Chr.4: 39552080-39552080 on Build GRCh38
rs776516228 Chr.4: 39552074-39552074 on Build GRCh38
rs771047548 Chr.4: 39552073-39552073 on Build GRCh38
rs373221151 Chr.4: 39552077-39552077 on Build GRCh38
rs112221868 Chr.4: 39552084-39552084 on Build GRCh38
rs1033720824 Chr.4: 39552052-39552052 on Build GRCh38
rs760543495 Chr.4: 39552096-39552096 on Build GRCh38
rs752553980 Chr.4: 39552205-39552206 on Build GRCh38
rs747531832 Chr.4: 39552200-39552200 on Build GRCh38
rs563289773 Chr.4: 39552089-39552089 on Build GRCh38
rs981108614 Chr.4: 39552049-39552049 on Build GRCh38
rs545690169 Chr.4: 39552188-39552188 on Build GRCh38

View Full Product Details