Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 29337023-29337296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP4
Entrez Gene ID
1846
Gene Name
dual specificity phosphatase 4
Gene Aliases
HVH2, MKP-2, MKP2, TYP
UniGene
Hs.417962
Gene Chromosome Location
Chr.8: 29333062-29350750 on Build GRCh38
Transcripts
NM_001394.6 NM_057158.3 XM_011544428.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5811223 c.778C>T p.R260W Chr.8: 29337160-29337160 on Build GRCh38
5501351 c.1085C>T p.S362L Chr.8: 29337126-29337126 on Build GRCh38
2789091 c.761G>T p.S254I Chr.8: 29337177-29337177 on Build GRCh38
1154987 c.884G>A p.S295N Chr.8: 29337054-29337054 on Build GRCh38
3432338 c.990C>T p.F330F Chr.8: 29337221-29337221 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753131743 Chr.8: 29337195-29337195 on Build GRCh38
rs776657488 Chr.8: 29337187-29337187 on Build GRCh38
rs749401971 Chr.8: 29337139-29337139 on Build GRCh38
rs747986111 Chr.8: 29337263-29337263 on Build GRCh38
rs748259910 Chr.8: 29337071-29337073 on Build GRCh38
rs767239485 Chr.8: 29337193-29337193 on Build GRCh38
rs370975967 Chr.8: 29337188-29337188 on Build GRCh38
rs7341662 Chr.8: 29337070-29337070 on Build GRCh38
rs962510069 Chr.8: 29337162-29337162 on Build GRCh38
rs374610674 Chr.8: 29337190-29337190 on Build GRCh38
rs368211989 Chr.8: 29337119-29337119 on Build GRCh38
rs749291210 Chr.8: 29337068-29337068 on Build GRCh38
rs758646705 Chr.8: 29337248-29337248 on Build GRCh38
rs747857516 Chr.8: 29337160-29337160 on Build GRCh38
rs547859194 Chr.8: 29337146-29337146 on Build GRCh38
rs772256235 Chr.8: 29337153-29337153 on Build GRCh38
rs773776161 Chr.8: 29337093-29337093 on Build GRCh38
rs772001516 Chr.8: 29337116-29337118 on Build GRCh38
rs771211136 Chr.8: 29337081-29337081 on Build GRCh38
rs777950416 Chr.8: 29337066-29337066 on Build GRCh38
rs138056315 Chr.8: 29337230-29337230 on Build GRCh38
rs151032013 Chr.8: 29337125-29337125 on Build GRCh38
rs772962826 Chr.8: 29337170-29337170 on Build GRCh38
rs7341661 Chr.8: 29337061-29337061 on Build GRCh38
rs769792484 Chr.8: 29337163-29337163 on Build GRCh38
rs542177995 Chr.8: 29337264-29337264 on Build GRCh38
rs150375319 Chr.8: 29337183-29337183 on Build GRCh38
rs974286158 Chr.8: 29337189-29337189 on Build GRCh38
rs745914444 Chr.8: 29337239-29337239 on Build GRCh38
rs376968600 Chr.8: 29337232-29337232 on Build GRCh38
rs769407772 Chr.8: 29337270-29337270 on Build GRCh38
rs372227576 Chr.8: 29337257-29337257 on Build GRCh38
rs754373118 Chr.8: 29337207-29337207 on Build GRCh38
rs919580482 Chr.8: 29337171-29337171 on Build GRCh38
rs768504501 Chr.8: 29337092-29337092 on Build GRCh38
rs370779406 Chr.8: 29337154-29337154 on Build GRCh38
rs764669713 Chr.8: 29337201-29337201 on Build GRCh38
rs147027609 Chr.8: 29337118-29337118 on Build GRCh38
rs368120060 Chr.8: 29337200-29337200 on Build GRCh38
rs199557082 Chr.8: 29337203-29337203 on Build GRCh38
rs754665752 Chr.8: 29337122-29337122 on Build GRCh38
rs756678785 Chr.8: 29337128-29337128 on Build GRCh38
rs1023257307 Chr.8: 29337105-29337105 on Build GRCh38
rs768045348 Chr.8: 29337124-29337124 on Build GRCh38
rs765050408 Chr.8: 29337098-29337098 on Build GRCh38
rs1015259596 Chr.8: 29337131-29337131 on Build GRCh38
rs778957038 Chr.8: 29337140-29337140 on Build GRCh38
rs773054971 Chr.8: 29337273-29337273 on Build GRCh38
rs757745526 Chr.8: 29337227-29337227 on Build GRCh38
rs912488794 Chr.8: 29337268-29337268 on Build GRCh38
rs749288021 Chr.8: 29337274-29337274 on Build GRCh38
rs761854287 Chr.8: 29337097-29337097 on Build GRCh38
rs552819707 Chr.8: 29337091-29337091 on Build GRCh38
rs374609431 Chr.8: 29337104-29337104 on Build GRCh38
rs953924603 Chr.8: 29337230-29337231 on Build GRCh38
rs774502089 Chr.8: 29337083-29337083 on Build GRCh38

View Full Product Details