Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 93068459-93068696 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP26C1
Entrez Gene ID
340665
Gene Name
cytochrome P450 family 26 subfamily C member 1
Gene Aliases
FFDD4
UniGene
Hs.369993
Gene Chromosome Location
Chr.10: 93061264-93068697 on Build GRCh38
Transcripts
NM_183374.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6420746 c.1496A>G p.H499R Chr.10: 93068624-93068624 on Build GRCh38
1675435 c.1391T>G p.L464R Chr.10: 93068519-93068519 on Build GRCh38
1202835 c.1456A>G p.T486A Chr.10: 93068584-93068584 on Build GRCh38
6500422 c.1541C>T p.S514L Chr.10: 93068669-93068669 on Build GRCh38
1191871 c.1342C>A p.H448N Chr.10: 93068470-93068470 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565717716 Chr.10: 93068599-93068599 on Build GRCh38
rs775168846 Chr.10: 93068511-93068511 on Build GRCh38
rs377145306 Chr.10: 93068657-93068657 on Build GRCh38
rs777493031 Chr.10: 93068563-93068563 on Build GRCh38
rs531070401 Chr.10: 93068656-93068656 on Build GRCh38
rs771586164 Chr.10: 93068595-93068595 on Build GRCh38
rs757100910 Chr.10: 93068640-93068640 on Build GRCh38
rs773498722 Chr.10: 93068607-93068607 on Build GRCh38
rs180969879 Chr.10: 93068620-93068620 on Build GRCh38
rs753971732 Chr.10: 93068635-93068635 on Build GRCh38
rs377151786 Chr.10: 93068522-93068522 on Build GRCh38
rs766860018 Chr.10: 93068525-93068525 on Build GRCh38
rs140502479 Chr.10: 93068619-93068619 on Build GRCh38
rs1029434604 Chr.10: 93068597-93068597 on Build GRCh38
rs372692848 Chr.10: 93068614-93068614 on Build GRCh38
rs369876110 Chr.10: 93068523-93068523 on Build GRCh38
rs745759230 Chr.10: 93068590-93068590 on Build GRCh38
rs767707524 Chr.10: 93068616-93068616 on Build GRCh38
rs1040487413 Chr.10: 93068674-93068674 on Build GRCh38
rs376961229 Chr.10: 93068596-93068596 on Build GRCh38
rs879940720 Chr.10: 93068601-93068601 on Build GRCh38
rs756334431 Chr.10: 93068561-93068561 on Build GRCh38
rs760518015 Chr.10: 93068528-93068528 on Build GRCh38
rs746731720 Chr.10: 93068598-93068598 on Build GRCh38
rs778944118 Chr.10: 93068583-93068583 on Build GRCh38
rs946933283 Chr.10: 93068603-93068603 on Build GRCh38
rs370290902 Chr.10: 93068610-93068610 on Build GRCh38
rs749254302 Chr.10: 93068497-93068497 on Build GRCh38
rs757186914 Chr.10: 93068570-93068570 on Build GRCh38
rs758329980 Chr.10: 93068658-93068658 on Build GRCh38
rs867332655 Chr.10: 93068605-93068605 on Build GRCh38
rs765585054 Chr.10: 93068516-93068516 on Build GRCh38
rs776377032 Chr.10: 93068667-93068667 on Build GRCh38
rs550466265 Chr.10: 93068514-93068514 on Build GRCh38
rs779598921 Chr.10: 93068659-93068659 on Build GRCh38
rs767984515 Chr.10: 93068521-93068558 on Build GRCh38
rs760060062 Chr.10: 93068615-93068615 on Build GRCh38
rs755273863 Chr.10: 93068479-93068479 on Build GRCh38
rs751728984 Chr.10: 93068527-93068527 on Build GRCh38
rs747704620 Chr.10: 93068675-93068675 on Build GRCh38
rs551889242 Chr.10: 93068537-93068537 on Build GRCh38
rs778859736 Chr.10: 93068609-93068609 on Build GRCh38
rs755171735 Chr.10: 93068564-93068564 on Build GRCh38
rs61729502 Chr.10: 93068529-93068529 on Build GRCh38
rs771337915 Chr.10: 93068677-93068677 on Build GRCh38
rs777614107 Chr.10: 93068494-93068494 on Build GRCh38
rs769763099 Chr.10: 93068489-93068489 on Build GRCh38
rs773752020 Chr.10: 93068501-93068501 on Build GRCh38
rs781525825 Chr.10: 93068484-93068484 on Build GRCh38
rs759002311 Chr.10: 93068509-93068509 on Build GRCh38
rs746500540 Chr.10: 93068665-93068665 on Build GRCh38
rs778518089 Chr.10: 93068644-93068644 on Build GRCh38
rs750732093 Chr.10: 93068604-93068604 on Build GRCh38

View Full Product Details