Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137716800-137717045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHMT1
Entrez Gene ID
79813
Gene Name
euchromatic histone lysine methyltransferase 1
Gene Aliases
EUHMTASE1, Eu-HMTase1, FP13812, GLP, GLP1, KMT1D, bA188C12.1
UniGene
Hs.495511
Gene Chromosome Location
Chr.9: 137618988-137836127 on Build GRCh38
Transcripts
NM_001145527.1 NM_024757.4 XM_005266105.4 XM_005266110.1 XM_006717288.2 XM_011519021.2 XM_011519022.2 XM_011519023.2 XM_011519024.2 XM_011519025.1 XM_011519026.2 XM_011519027.2 XM_011519028.2 XM_011519033.2 XM_017015134.1 XM_017015135.1 XM_017015136.1 XM_017015137.1 XM_017015138.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7366006 c.478G>T p.G160C Chr.9: 137717018-137717018 on Build GRCh38
1598174 c.337G>A p.D113N Chr.9: 137716877-137716877 on Build GRCh38
7366007 c.385G>T p.G129C Chr.9: 137717018-137717018 on Build GRCh38
1107165 c.176G>A p.R59Q Chr.9: 137716809-137716809 on Build GRCh38
1598176 c.269G>A p.R90Q Chr.9: 137716809-137716809 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924620448 Chr.9: 137716927-137716927 on Build GRCh38
rs781024598 Chr.9: 137716848-137716848 on Build GRCh38
rs562411087 Chr.9: 137716928-137716928 on Build GRCh38
rs772079296 Chr.9: 137716855-137716855 on Build GRCh38
rs775347185 Chr.9: 137717024-137717024 on Build GRCh38
rs758617464 Chr.9: 137716937-137716937 on Build GRCh38
rs1003872402 Chr.9: 137717003-137717003 on Build GRCh38
rs138824805 Chr.9: 137716849-137716849 on Build GRCh38
rs369562168 Chr.9: 137716850-137716850 on Build GRCh38
rs751483939 Chr.9: 137716875-137716875 on Build GRCh38
rs370834767 Chr.9: 137716994-137716994 on Build GRCh38
rs200982880 Chr.9: 137716945-137716945 on Build GRCh38
rs139431280 Chr.9: 137716883-137716883 on Build GRCh38
rs770086341 Chr.9: 137716909-137716909 on Build GRCh38
rs3812497 Chr.9: 137716984-137716984 on Build GRCh38
rs140387451 Chr.9: 137716910-137716910 on Build GRCh38
rs151008340 Chr.9: 137716978-137716978 on Build GRCh38
rs571203834 Chr.9: 137716830-137716830 on Build GRCh38
rs746186916 Chr.9: 137716908-137716908 on Build GRCh38
rs559706048 Chr.9: 137716870-137716870 on Build GRCh38
rs199913017 Chr.9: 137717006-137717006 on Build GRCh38
rs898224027 Chr.9: 137717014-137717014 on Build GRCh38
rs377575553 Chr.9: 137716991-137716991 on Build GRCh38
rs763653989 Chr.9: 137716915-137716915 on Build GRCh38
rs779153176 Chr.9: 137716885-137716885 on Build GRCh38
rs780621678 Chr.9: 137717022-137717022 on Build GRCh38
rs752755202 Chr.9: 137716877-137716877 on Build GRCh38
rs757024199 Chr.9: 137716839-137716839 on Build GRCh38
rs373407070 Chr.9: 137716967-137716967 on Build GRCh38
rs142199482 Chr.9: 137716851-137716851 on Build GRCh38
rs199703555 Chr.9: 137716865-137716865 on Build GRCh38
rs902223841 Chr.9: 137716958-137716958 on Build GRCh38
rs147648583 Chr.9: 137716869-137716869 on Build GRCh38
rs761269044 Chr.9: 137716981-137716981 on Build GRCh38
rs374844911 Chr.9: 137716890-137716890 on Build GRCh38
rs781479211 Chr.9: 137717011-137717011 on Build GRCh38
rs368961311 Chr.9: 137716873-137716873 on Build GRCh38
rs142652443 Chr.9: 137716977-137716977 on Build GRCh38
rs147764684 Chr.9: 137716947-137716947 on Build GRCh38
rs554656861 Chr.9: 137716861-137716861 on Build GRCh38
rs797045043 Chr.9: 137716931-137716931 on Build GRCh38
rs866531374 Chr.9: 137716946-137716946 on Build GRCh38
rs763562241 Chr.9: 137716874-137716874 on Build GRCh38
rs144352877 Chr.9: 137716903-137716903 on Build GRCh38
rs139461232 Chr.9: 137716972-137716972 on Build GRCh38
rs773781896 Chr.9: 137716916-137716916 on Build GRCh38
rs747517245 Chr.9: 137716900-137716900 on Build GRCh38
rs773115118 Chr.9: 137716985-137716985 on Build GRCh38
rs766195455 Chr.9: 137716988-137716988 on Build GRCh38
rs757250849 Chr.9: 137716876-137716876 on Build GRCh38
rs576731460 Chr.9: 137716852-137716852 on Build GRCh38
rs751633207 Chr.9: 137716942-137716942 on Build GRCh38
rs769036698 Chr.9: 137717025-137717025 on Build GRCh38
rs771499811 Chr.9: 137716901-137716901 on Build GRCh38
rs144323841 Chr.9: 137716930-137716930 on Build GRCh38
rs540121859 Chr.9: 137716838-137716838 on Build GRCh38
rs372415033 Chr.9: 137716941-137716941 on Build GRCh38
rs150135875 Chr.9: 137717020-137717020 on Build GRCh38
rs1046702208 Chr.9: 137716943-137716943 on Build GRCh38
rs762570084 Chr.9: 137716913-137716913 on Build GRCh38
rs764884859 Chr.9: 137716872-137716872 on Build GRCh38
rs767390279 Chr.9: 137716929-137716929 on Build GRCh38
rs369253537 Chr.9: 137716898-137716898 on Build GRCh38
rs886740560 Chr.9: 137716858-137716858 on Build GRCh38
rs144603232 Chr.9: 137716856-137716856 on Build GRCh38
rs766141140 Chr.9: 137716933-137716933 on Build GRCh38
rs867311907 Chr.9: 137716867-137716867 on Build GRCh38
rs752954008 Chr.9: 137716936-137716936 on Build GRCh38
rs142271310 Chr.9: 137716894-137716894 on Build GRCh38
rs780324829 Chr.9: 137716837-137716842 on Build GRCh38
rs990229301 Chr.9: 137716931-137716931 on Build GRCh38
rs574402576 Chr.9: 137716864-137716864 on Build GRCh38
rs753513313 Chr.9: 137716990-137716990 on Build GRCh38
rs760591719 Chr.9: 137716987-137716987 on Build GRCh38
rs374172429 Chr.9: 137716997-137716997 on Build GRCh38
rs750935706 Chr.9: 137717012-137717012 on Build GRCh38
rs797044709 Chr.9: 137716838-137716839 on Build GRCh38

View Full Product Details