Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 4125173-4125446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf201
Entrez Gene ID
404220
Gene Name
chromosome 6 open reading frame 201
Gene Aliases
dJ1013A10.5
UniGene
Hs.716731
Gene Chromosome Location
Chr.6: 4079197-4130788 on Build GRCh38
Transcripts
NM_001085401.2 NR_104463.1 NR_104464.1 XM_011514613.2 XM_017010861.1 NM_001166010.1 NM_006117.2 NM_206836.2 NR_028588.1 XM_006714957.1 XM_017010170.1

Gene Symbol
ECI2
Entrez Gene ID
10455
Gene Name
enoyl-CoA delta isomerase 2
Gene Aliases
ACBD2, DRS-1, DRS1, HCA88, PECI, dJ1013A10.3
UniGene
Hs.15250
Gene Chromosome Location
Chr.6: 4115693-4135597 on Build GRCh38
Transcripts
NM_001085401.2 NR_104463.1 NR_104464.1 XM_011514613.2 XM_017010861.1 NM_001166010.1 NM_006117.2 NM_206836.2 NR_028588.1 XM_006714957.1 XM_017010170.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5479472 c.772G>T p.D258Y Chr.6: 4125273-4125273 on Build GRCh38
3430658 c.771C>T p.F257F Chr.6: 4125274-4125274 on Build GRCh38
6616441 c.585-6C>A p.? Chr.6: 4125376-4125376 on Build GRCh38
5466856 c.771C>A p.F257L Chr.6: 4125274-4125274 on Build GRCh38
7215075 c.705+10C>T p.? Chr.6: 4125240-4125240 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376441298 Chr.6: 4125209-4125209 on Build GRCh38
rs937249944 Chr.6: 4125322-4125322 on Build GRCh38
rs367695337 Chr.6: 4125268-4125268 on Build GRCh38
rs746110782 Chr.6: 4125419-4125419 on Build GRCh38
rs779380039 Chr.6: 4125414-4125414 on Build GRCh38
rs376485977 Chr.6: 4125208-4125208 on Build GRCh38
rs761538340 Chr.6: 4125412-4125422 on Build GRCh38
rs748636273 Chr.6: 4125224-4125224 on Build GRCh38
rs142672246 Chr.6: 4125334-4125334 on Build GRCh38
rs751307625 Chr.6: 4125426-4125426 on Build GRCh38
rs763108114 Chr.6: 4125245-4125245 on Build GRCh38
rs985158395 Chr.6: 4125238-4125238 on Build GRCh38
rs756584624 Chr.6: 4125384-4125384 on Build GRCh38
rs200235549 Chr.6: 4125363-4125363 on Build GRCh38
rs779511364 Chr.6: 4125287-4125287 on Build GRCh38
rs542728134 Chr.6: 4125226-4125226 on Build GRCh38
rs192626144 Chr.6: 4125380-4125380 on Build GRCh38
rs752638238 Chr.6: 4125335-4125335 on Build GRCh38
rs756881420 Chr.6: 4125206-4125206 on Build GRCh38
rs774680876 Chr.6: 4125308-4125308 on Build GRCh38
rs764921233 Chr.6: 4125366-4125366 on Build GRCh38
rs765009995 Chr.6: 4125274-4125274 on Build GRCh38
rs757679715 Chr.6: 4125398-4125398 on Build GRCh38
rs11751668 Chr.6: 4125240-4125240 on Build GRCh38
rs778015184 Chr.6: 4125294-4125294 on Build GRCh38
rs771185761 Chr.6: 4125299-4125299 on Build GRCh38
rs41302847 Chr.6: 4125267-4125267 on Build GRCh38
rs549462603 Chr.6: 4125347-4125347 on Build GRCh38
rs659305 Chr.6: 4125421-4125421 on Build GRCh38
rs758007119 Chr.6: 4125376-4125376 on Build GRCh38
rs775659179 Chr.6: 4125336-4125336 on Build GRCh38
rs913946924 Chr.6: 4125310-4125310 on Build GRCh38
rs756861928 Chr.6: 4125273-4125273 on Build GRCh38
rs759745663 Chr.6: 4125249-4125249 on Build GRCh38
rs750065674 Chr.6: 4125276-4125276 on Build GRCh38
rs1802393 Chr.6: 4125352-4125352 on Build GRCh38
rs549800477 Chr.6: 4125291-4125291 on Build GRCh38
rs751014667 Chr.6: 4125382-4125382 on Build GRCh38
rs867251171 Chr.6: 4125277-4125277 on Build GRCh38
rs766762131 Chr.6: 4125372-4125372 on Build GRCh38
rs774430756 Chr.6: 4125247-4125247 on Build GRCh38
rs944517561 Chr.6: 4125381-4125381 on Build GRCh38
rs202185765 Chr.6: 4125356-4125356 on Build GRCh38
rs373210025 Chr.6: 4125207-4125207 on Build GRCh38
rs757956244 Chr.6: 4125285-4125285 on Build GRCh38
rs148485246 Chr.6: 4125317-4125317 on Build GRCh38
rs778400322 Chr.6: 4125386-4125386 on Build GRCh38
rs145542207 Chr.6: 4125252-4125252 on Build GRCh38
rs144214404 Chr.6: 4125269-4125269 on Build GRCh38
rs748685734 Chr.6: 4125288-4125288 on Build GRCh38
rs139545999 Chr.6: 4125259-4125259 on Build GRCh38
rs749735565 Chr.6: 4125390-4125390 on Build GRCh38
rs749602104 Chr.6: 4125295-4125295 on Build GRCh38
rs893128818 Chr.6: 4125339-4125339 on Build GRCh38
rs935924883 Chr.6: 4125353-4125353 on Build GRCh38
rs370913419 Chr.6: 4125210-4125210 on Build GRCh38
rs751319560 Chr.6: 4125252-4125252 on Build GRCh38
rs764022867 Chr.6: 4125350-4125350 on Build GRCh38

View Full Product Details