Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 34868683-34868926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLGAP3
Entrez Gene ID
58512
Gene Name
DLG associated protein 3
Gene Aliases
DAP3, SAPAP3
UniGene
Hs.733945
Gene Chromosome Location
Chr.1: 34862120-34929585 on Build GRCh38
Transcripts
NM_001080418.2 XM_011541879.2 XM_011541880.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3488840 c.2322G>A p.W774* Chr.1: 34868768-34868768 on Build GRCh38
3488839 c.2366C>T p.S789F Chr.1: 34868724-34868724 on Build GRCh38
5587628 c.2296C>T p.P766S Chr.1: 34868794-34868794 on Build GRCh38
6856499 c.2169delC p.T724fs*52 Chr.1: 34868921-34868921 on Build GRCh38
1203756 c.2269G>T p.G757C Chr.1: 34868821-34868821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542950831 Chr.1: 34868870-34868870 on Build GRCh38
rs761387951 Chr.1: 34868844-34868844 on Build GRCh38
rs928166467 Chr.1: 34868831-34868831 on Build GRCh38
rs773331125 Chr.1: 34868705-34868705 on Build GRCh38
rs369099002 Chr.1: 34868731-34868731 on Build GRCh38
rs763155158 Chr.1: 34868774-34868774 on Build GRCh38
rs749399902 Chr.1: 34868821-34868821 on Build GRCh38
rs745644846 Chr.1: 34868751-34868751 on Build GRCh38
rs948963751 Chr.1: 34868764-34868764 on Build GRCh38
rs750007956 Chr.1: 34868801-34868801 on Build GRCh38
rs759780621 Chr.1: 34868825-34868825 on Build GRCh38
rs746903257 Chr.1: 34868760-34868760 on Build GRCh38
rs772418605 Chr.1: 34868904-34868904 on Build GRCh38
rs376879114 Chr.1: 34868762-34868762 on Build GRCh38
rs747997191 Chr.1: 34868881-34868881 on Build GRCh38
rs916821098 Chr.1: 34868841-34868841 on Build GRCh38
rs150088046 Chr.1: 34868708-34868708 on Build GRCh38
rs148167929 Chr.1: 34868759-34868759 on Build GRCh38
rs1027736188 Chr.1: 34868793-34868793 on Build GRCh38
rs746132929 Chr.1: 34868901-34868901 on Build GRCh38
rs77048244 Chr.1: 34868711-34868711 on Build GRCh38
rs774657338 Chr.1: 34868779-34868779 on Build GRCh38
rs753276072 Chr.1: 34868785-34868785 on Build GRCh38
rs768153608 Chr.1: 34868727-34868727 on Build GRCh38
rs754237871 Chr.1: 34868843-34868843 on Build GRCh38
rs750127207 Chr.1: 34868852-34868852 on Build GRCh38
rs142976280 Chr.1: 34868905-34868905 on Build GRCh38
rs372616310 Chr.1: 34868761-34868761 on Build GRCh38
rs759589416 Chr.1: 34868766-34868766 on Build GRCh38
rs572015126 Chr.1: 34868822-34868822 on Build GRCh38
rs761630237 Chr.1: 34868797-34868808 on Build GRCh38
rs778636829 Chr.1: 34868746-34868746 on Build GRCh38
rs771067661 Chr.1: 34868891-34868891 on Build GRCh38
rs776159291 Chr.1: 34868710-34868711 on Build GRCh38
rs11587343 Chr.1: 34868849-34868849 on Build GRCh38
rs373832741 Chr.1: 34868833-34868833 on Build GRCh38
rs765322873 Chr.1: 34868704-34868704 on Build GRCh38
rs771794556 Chr.1: 34868752-34868752 on Build GRCh38
rs758215471 Chr.1: 34868803-34868803 on Build GRCh38
rs763551958 Chr.1: 34868797-34868797 on Build GRCh38
rs771172185 Chr.1: 34868778-34868778 on Build GRCh38
rs878931421 Chr.1: 34868809-34868809 on Build GRCh38
rs761151648 Chr.1: 34868840-34868840 on Build GRCh38
rs201877472 Chr.1: 34868796-34868796 on Build GRCh38
rs779804155 Chr.1: 34868804-34868804 on Build GRCh38
rs767955367 Chr.1: 34868782-34868782 on Build GRCh38
rs150619481 Chr.1: 34868900-34868900 on Build GRCh38
rs541046831 Chr.1: 34868786-34868786 on Build GRCh38
rs753189951 Chr.1: 34868728-34868728 on Build GRCh38
rs768638422 Chr.1: 34868812-34868812 on Build GRCh38
rs201441073 Chr.1: 34868798-34868798 on Build GRCh38
rs765300605 Chr.1: 34868781-34868781 on Build GRCh38
rs149415698 Chr.1: 34868721-34868721 on Build GRCh38
rs200442780 Chr.1: 34868707-34868707 on Build GRCh38
rs774747027 Chr.1: 34868823-34868823 on Build GRCh38
rs776466719 Chr.1: 34868730-34868730 on Build GRCh38
rs781194450 Chr.1: 34868871-34868871 on Build GRCh38
rs746992705 Chr.1: 34868810-34868810 on Build GRCh38
rs370934625 Chr.1: 34868853-34868853 on Build GRCh38
rs755715413 Chr.1: 34868729-34868729 on Build GRCh38
rs755600491 Chr.1: 34868713-34868713 on Build GRCh38
rs373836246 Chr.1: 34868741-34868741 on Build GRCh38
rs997762298 Chr.1: 34868792-34868792 on Build GRCh38
rs778639162 Chr.1: 34868800-34868800 on Build GRCh38
rs762566448 Chr.1: 34868834-34868834 on Build GRCh38
rs777773426 Chr.1: 34868814-34868814 on Build GRCh38
rs779894036 Chr.1: 34868854-34868854 on Build GRCh38

View Full Product Details