Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 38825626-38825897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX57
Entrez Gene ID
90957
Gene Name
DEAH-box helicase 57
Gene Aliases
DDX57
UniGene
Hs.468226
Gene Chromosome Location
Chr.2: 38797729-38875883 on Build GRCh38
Transcripts
NM_198963.1 XM_011533154.2 XM_011533155.2 XM_011533156.2 XM_011533157.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4133835 c.2985A>C p.Q995H Chr.2: 38825876-38825876 on Build GRCh38
4415479 c.2995T>C p.L999L Chr.2: 38825866-38825866 on Build GRCh38
5697533 c.2988A>G p.R996R Chr.2: 38825873-38825873 on Build GRCh38
1020839 c.2992C>T p.P998S Chr.2: 38825869-38825869 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs999556333 Chr.2: 38825798-38825798 on Build GRCh38
rs770156325 Chr.2: 38825836-38825836 on Build GRCh38
rs775573056 Chr.2: 38825829-38825830 on Build GRCh38
rs753496198 Chr.2: 38825796-38825796 on Build GRCh38
rs948637718 Chr.2: 38825773-38825773 on Build GRCh38
rs938756244 Chr.2: 38825747-38825747 on Build GRCh38
rs930457208 Chr.2: 38825754-38825754 on Build GRCh38
rs778502834 Chr.2: 38825819-38825819 on Build GRCh38
rs771064623 Chr.2: 38825844-38825844 on Build GRCh38
rs75545406 Chr.2: 38825664-38825664 on Build GRCh38
rs774398421 Chr.2: 38825847-38825847 on Build GRCh38
rs549326932 Chr.2: 38825866-38825866 on Build GRCh38
rs182658727 Chr.2: 38825762-38825762 on Build GRCh38
rs749444248 Chr.2: 38825839-38825839 on Build GRCh38
rs762356068 Chr.2: 38825737-38825737 on Build GRCh38
rs772349404 Chr.2: 38825830-38825830 on Build GRCh38
rs747589057 Chr.2: 38825821-38825821 on Build GRCh38
rs539371415 Chr.2: 38825801-38825801 on Build GRCh38
rs919039599 Chr.2: 38825750-38825750 on Build GRCh38
rs761744689 Chr.2: 38825864-38825865 on Build GRCh38
rs951450590 Chr.2: 38825684-38825684 on Build GRCh38
rs1045687680 Chr.2: 38825807-38825807 on Build GRCh38
rs773436216 Chr.2: 38825838-38825838 on Build GRCh38
rs915749992 Chr.2: 38825767-38825767 on Build GRCh38
rs982843452 Chr.2: 38825703-38825703 on Build GRCh38
rs748430646 Chr.2: 38825825-38825825 on Build GRCh38
rs531043285 Chr.2: 38825857-38825857 on Build GRCh38
rs916102094 Chr.2: 38825759-38825759 on Build GRCh38
rs903471114 Chr.2: 38825775-38825775 on Build GRCh38
rs533145461 Chr.2: 38825668-38825668 on Build GRCh38
rs759465409 Chr.2: 38825816-38825818 on Build GRCh38
rs144335881 Chr.2: 38825741-38825741 on Build GRCh38
rs1018074559 Chr.2: 38825650-38825650 on Build GRCh38
rs865915191 Chr.2: 38825863-38825864 on Build GRCh38
rs117517065 Chr.2: 38825653-38825653 on Build GRCh38
rs760412234 Chr.2: 38825868-38825868 on Build GRCh38
rs965684580 Chr.2: 38825718-38825718 on Build GRCh38
rs781658943 Chr.2: 38825822-38825822 on Build GRCh38
rs750747848 Chr.2: 38825830-38825830 on Build GRCh38

View Full Product Details