Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 13700449-13700716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH5
Entrez Gene ID
1767
Gene Name
dynein axonemal heavy chain 5
Gene Aliases
CILD3, DNAHC5, HL1, KTGNR, PCD
UniGene
Hs.212360
Gene Chromosome Location
Chr.5: 13690328-14011829 on Build GRCh38
Transcripts
NM_001369.2 XM_005248262.3 XM_017009177.1 XM_017009178.1 XM_017009179.1 XM_017009185.1 XM_017009186.1 XM_017009188.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1486305 c.13682T>A p.F4561Y Chr.5: 13700681-13700681 on Build GRCh38
6646645 c.13723+2T>C p.? Chr.5: 13700638-13700638 on Build GRCh38
3608779 c.13704G>A p.R4568R Chr.5: 13700659-13700659 on Build GRCh38
4141423 c.13664C>G p.S4555* Chr.5: 13700699-13700699 on Build GRCh38
7092080 c.13696G>C p.V4566L Chr.5: 13700667-13700667 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1026067081 Chr.5: 13700535-13700535 on Build GRCh38
rs923901117 Chr.5: 13700498-13700498 on Build GRCh38
rs954379608 Chr.5: 13700685-13700685 on Build GRCh38
rs771284437 Chr.5: 13700600-13700600 on Build GRCh38
rs762303341 Chr.5: 13700521-13700521 on Build GRCh38
rs747291233 Chr.5: 13700644-13700644 on Build GRCh38
rs776530999 Chr.5: 13700659-13700659 on Build GRCh38
rs939271838 Chr.5: 13700517-13700517 on Build GRCh38
rs772147945 Chr.5: 13700616-13700616 on Build GRCh38
rs374221053 Chr.5: 13700674-13700674 on Build GRCh38
rs200891691 Chr.5: 13700667-13700667 on Build GRCh38
rs751090601 Chr.5: 13700688-13700688 on Build GRCh38
rs371251694 Chr.5: 13700529-13700529 on Build GRCh38
rs536407240 Chr.5: 13700599-13700599 on Build GRCh38
rs963291171 Chr.5: 13700504-13700504 on Build GRCh38
rs761546028 Chr.5: 13700664-13700664 on Build GRCh38
rs947808960 Chr.5: 13700588-13700588 on Build GRCh38
rs765032639 Chr.5: 13700666-13700666 on Build GRCh38
rs766044681 Chr.5: 13700677-13700677 on Build GRCh38
rs779305798 Chr.5: 13700603-13700603 on Build GRCh38
rs569149019 Chr.5: 13700578-13700578 on Build GRCh38
rs74377312 Chr.5: 13700618-13700618 on Build GRCh38
rs867555208 Chr.5: 13700570-13700570 on Build GRCh38
rs35809147 Chr.5: 13700512-13700513 on Build GRCh38
rs768846678 Chr.5: 13700649-13700649 on Build GRCh38
rs79782251 Chr.5: 13700609-13700609 on Build GRCh38
rs916416098 Chr.5: 13700583-13700583 on Build GRCh38
rs778329162 Chr.5: 13700589-13700589 on Build GRCh38

View Full Product Details