Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 177436167-177436440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGA
Entrez Gene ID
175
Gene Name
aspartylglucosaminidase
Gene Aliases
AGU, ASRG, GA
UniGene
Hs.207776
Gene Chromosome Location
Chr.4: 177430774-177442503 on Build GRCh38
Transcripts
NM_000027.3 NM_001171988.1 NR_033655.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
390932 c.654T>G p.I218M Chr.4: 177436320-177436320 on Build GRCh38
7354712 c.623G>T p.G208V Chr.4: 177436351-177436351 on Build GRCh38
6323851 c.656C>T p.A219V Chr.4: 177436318-177436318 on Build GRCh38
7416112 c.655G>T p.A219S Chr.4: 177436319-177436319 on Build GRCh38
4774700 c.623-10delT p.? Chr.4: 177436361-177436361 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373612365 Chr.4: 177436354-177436354 on Build GRCh38
rs779019286 Chr.4: 177436361-177436362 on Build GRCh38
rs768636689 Chr.4: 177436316-177436316 on Build GRCh38
rs772311291 Chr.4: 177436360-177436361 on Build GRCh38
rs113118047 Chr.4: 177436193-177436193 on Build GRCh38
rs377145157 Chr.4: 177436242-177436242 on Build GRCh38
rs888352005 Chr.4: 177436355-177436355 on Build GRCh38
rs756908895 Chr.4: 177436266-177436266 on Build GRCh38
rs961216952 Chr.4: 177436408-177436408 on Build GRCh38
rs386833431 Chr.4: 177436297-177436297 on Build GRCh38
rs767924247 Chr.4: 177436336-177436336 on Build GRCh38
rs778434616 Chr.4: 177436364-177436364 on Build GRCh38
rs771926983 Chr.4: 177436296-177436296 on Build GRCh38
rs758111896 Chr.4: 177436384-177436384 on Build GRCh38
rs759848605 Chr.4: 177436228-177436228 on Build GRCh38
rs763070208 Chr.4: 177436325-177436325 on Build GRCh38
rs774485932 Chr.4: 177436330-177436330 on Build GRCh38
rs768012132 Chr.4: 177436241-177436241 on Build GRCh38
rs778579846 Chr.4: 177436285-177436285 on Build GRCh38
rs201125635 Chr.4: 177436358-177436358 on Build GRCh38
rs886059261 Chr.4: 177436317-177436317 on Build GRCh38
rs753356588 Chr.4: 177436256-177436256 on Build GRCh38
rs747696754 Chr.4: 177436241-177436242 on Build GRCh38
rs79838824 Chr.4: 177436212-177436212 on Build GRCh38
rs571447058 Chr.4: 177436196-177436196 on Build GRCh38
rs71599314 Chr.4: 177436412-177436413 on Build GRCh38
rs879762179 Chr.4: 177436386-177436386 on Build GRCh38
rs769278659 Chr.4: 177436219-177436219 on Build GRCh38
rs750039624 Chr.4: 177436368-177436368 on Build GRCh38
rs386402426 Chr.4: 177436413-177436414 on Build GRCh38
rs1057517175 Chr.4: 177436275-177436275 on Build GRCh38
rs773208051 Chr.4: 177436324-177436324 on Build GRCh38
rs1052361578 Chr.4: 177436342-177436342 on Build GRCh38
rs745896951 Chr.4: 177436361-177436361 on Build GRCh38
rs1053273669 Chr.4: 177436210-177436210 on Build GRCh38
rs753309242 Chr.4: 177436357-177436357 on Build GRCh38
rs201441868 Chr.4: 177436329-177436329 on Build GRCh38
rs780063830 Chr.4: 177436385-177436385 on Build GRCh38
rs1039067399 Chr.4: 177436306-177436306 on Build GRCh38
rs369432133 Chr.4: 177436309-177436309 on Build GRCh38
rs935244358 Chr.4: 177436237-177436237 on Build GRCh38
rs753052665 Chr.4: 177436341-177436341 on Build GRCh38
rs756597510 Chr.4: 177436249-177436249 on Build GRCh38
rs138699617 Chr.4: 177436299-177436299 on Build GRCh38
rs551697415 Chr.4: 177436215-177436215 on Build GRCh38
rs759903678 Chr.4: 177436333-177436333 on Build GRCh38
rs923840516 Chr.4: 177436216-177436216 on Build GRCh38
rs745689169 Chr.4: 177436287-177436287 on Build GRCh38
rs947410301 Chr.4: 177436304-177436304 on Build GRCh38
rs933465949 Chr.4: 177436317-177436317 on Build GRCh38
rs761124709 Chr.4: 177436346-177436346 on Build GRCh38

View Full Product Details