Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54174586-54174857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5RL
Entrez Gene ID
606495
Gene Name
cytochrome b5 reductase like
Gene Aliases
-
UniGene
Hs.591426
Gene Chromosome Location
Chr.1: 54172336-54200073 on Build GRCh38
Transcripts
NM_001031672.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4919815 c.848A>G p.K283R Chr.1: 54174719-54174719 on Build GRCh38
4885258 c.925G>T p.E309* Chr.1: 54174642-54174642 on Build GRCh38
5086532 c.931T>G p.S311A Chr.1: 54174636-54174636 on Build GRCh38
3985143 c.845G>A p.R282K Chr.1: 54174722-54174722 on Build GRCh38
6638766 c.802C>A p.L268M Chr.1: 54174765-54174765 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201353692 Chr.1: 54174753-54174753 on Build GRCh38
rs780279305 Chr.1: 54174715-54174715 on Build GRCh38
rs61741270 Chr.1: 54174655-54174655 on Build GRCh38
rs759161836 Chr.1: 54174728-54174728 on Build GRCh38
rs190838484 Chr.1: 54174818-54174818 on Build GRCh38
rs763985375 Chr.1: 54174756-54174756 on Build GRCh38
rs781522510 Chr.1: 54174607-54174607 on Build GRCh38
rs773462054 Chr.1: 54174726-54174726 on Build GRCh38
rs780876546 Chr.1: 54174711-54174711 on Build GRCh38
rs752188035 Chr.1: 54174744-54174744 on Build GRCh38
rs530303527 Chr.1: 54174712-54174712 on Build GRCh38
rs375961783 Chr.1: 54174613-54174613 on Build GRCh38
rs758325962 Chr.1: 54174694-54174694 on Build GRCh38
rs756349100 Chr.1: 54174798-54174798 on Build GRCh38
rs753489709 Chr.1: 54174657-54174657 on Build GRCh38
rs750255497 Chr.1: 54174776-54174776 on Build GRCh38
rs755737482 Chr.1: 54174692-54174692 on Build GRCh38
rs373694357 Chr.1: 54174765-54174765 on Build GRCh38
rs759133773 Chr.1: 54174672-54174672 on Build GRCh38
rs752455211 Chr.1: 54174691-54174691 on Build GRCh38
rs764934556 Chr.1: 54174743-54174743 on Build GRCh38
rs757112805 Chr.1: 54174702-54174702 on Build GRCh38
rs762288981 Chr.1: 54174654-54174654 on Build GRCh38
rs867559516 Chr.1: 54174777-54174777 on Build GRCh38
rs769265229 Chr.1: 54174719-54174719 on Build GRCh38
rs186035697 Chr.1: 54174674-54174674 on Build GRCh38
rs763937870 Chr.1: 54174616-54174616 on Build GRCh38
rs751470173 Chr.1: 54174760-54174760 on Build GRCh38
rs763491587 Chr.1: 54174640-54174640 on Build GRCh38
rs199663086 Chr.1: 54174718-54174718 on Build GRCh38
rs367565690 Chr.1: 54174800-54174800 on Build GRCh38
rs775730518 Chr.1: 54174638-54174638 on Build GRCh38
rs751330524 Chr.1: 54174695-54174695 on Build GRCh38
rs762259188 Chr.1: 54174723-54174723 on Build GRCh38
rs772433235 Chr.1: 54174725-54174725 on Build GRCh38
rs368711832 Chr.1: 54174681-54174681 on Build GRCh38
rs774845290 Chr.1: 54174652-54174652 on Build GRCh38
rs181944525 Chr.1: 54174834-54174834 on Build GRCh38
rs755071358 Chr.1: 54174826-54174826 on Build GRCh38
rs577570807 Chr.1: 54174757-54174757 on Build GRCh38
rs767280736 Chr.1: 54174772-54174772 on Build GRCh38
rs769874126 Chr.1: 54174623-54174623 on Build GRCh38
rs61741274 Chr.1: 54174645-54174645 on Build GRCh38
rs377135102 Chr.1: 54174628-54174628 on Build GRCh38

View Full Product Details