Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 1027786-1028041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERC1
Entrez Gene ID
23085
Gene Name
ELKS/RAB6-interacting/CAST family member 1
Gene Aliases
Cast2, ELKS, ERC-1, RAB6IP2
UniGene
Hs.601216
Gene Chromosome Location
Chr.12: 990010-1495933 on Build GRCh38
Transcripts
NM_001301248.1 NM_178039.3 NM_178040.3 NR_027946.2 NR_027948.2 NR_027949.2 XM_011520941.2 XM_017019052.1 XM_017019053.1 XM_017019054.1 XM_017019055.1 XM_017019056.1 XM_017019057.1 XM_017019058.1 XM_017019059.1 XM_017019060.1 XM_017019061.1 XM_017019062.1 XM_017019063.1 XM_017019064.1 XM_017019065.1 XM_017019066.1 XM_017019067.1 XM_017019068.1 XM_017019069.1 XM_017019070.1 XM_017019071.1 XM_017019072.1 XM_017019073.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6071363 c.120G>T p.G40G Chr.12: 1028023-1028023 on Build GRCh38
935409 c.100C>T p.R34* Chr.12: 1028003-1028003 on Build GRCh38
4038980 c.45C>T p.S15S Chr.12: 1027948-1027948 on Build GRCh38
225325 c.20C>T p.S7F Chr.12: 1027923-1027923 on Build GRCh38
691994 c.97C>T p.R33C Chr.12: 1028000-1028000 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763176873 Chr.12: 1027934-1027934 on Build GRCh38
rs199989730 Chr.12: 1027884-1027884 on Build GRCh38
rs61740169 Chr.12: 1027951-1027951 on Build GRCh38
rs377032908 Chr.12: 1027961-1027961 on Build GRCh38
rs73606389 Chr.12: 1027817-1027817 on Build GRCh38
rs755733420 Chr.12: 1027881-1027881 on Build GRCh38
rs753375786 Chr.12: 1027883-1027883 on Build GRCh38
rs369773653 Chr.12: 1027919-1027919 on Build GRCh38
rs374326925 Chr.12: 1028017-1028017 on Build GRCh38
rs774585775 Chr.12: 1028003-1028003 on Build GRCh38
rs754782634 Chr.12: 1027989-1027989 on Build GRCh38
rs775535153 Chr.12: 1027979-1027979 on Build GRCh38
rs745911310 Chr.12: 1028004-1028004 on Build GRCh38
rs34283065 Chr.12: 1027956-1027956 on Build GRCh38
rs992152183 Chr.12: 1027851-1027852 on Build GRCh38
rs549963000 Chr.12: 1027962-1027962 on Build GRCh38
rs936122168 Chr.12: 1027916-1027916 on Build GRCh38
rs771721336 Chr.12: 1027861-1027861 on Build GRCh38
rs778200446 Chr.12: 1027991-1027991 on Build GRCh38
rs764425580 Chr.12: 1028022-1028022 on Build GRCh38
rs567169596 Chr.12: 1027854-1027854 on Build GRCh38
rs779729714 Chr.12: 1027915-1027915 on Build GRCh38
rs374468019 Chr.12: 1027926-1027926 on Build GRCh38
rs749881506 Chr.12: 1027967-1027967 on Build GRCh38
rs144267759 Chr.12: 1027882-1027882 on Build GRCh38
rs776030209 Chr.12: 1028016-1028016 on Build GRCh38
rs771983448 Chr.12: 1027908-1027908 on Build GRCh38
rs745804239 Chr.12: 1027897-1027897 on Build GRCh38
rs886848143 Chr.12: 1027980-1027980 on Build GRCh38
rs571730696 Chr.12: 1028000-1028000 on Build GRCh38
rs746820047 Chr.12: 1027984-1027984 on Build GRCh38
rs771123374 Chr.12: 1027941-1027941 on Build GRCh38
rs150005389 Chr.12: 1027831-1027831 on Build GRCh38
rs943079739 Chr.12: 1027842-1027842 on Build GRCh38
rs761056838 Chr.12: 1027953-1027953 on Build GRCh38
rs778867282 Chr.12: 1027885-1027885 on Build GRCh38
rs143595866 Chr.12: 1027852-1027854 on Build GRCh38
rs979345548 Chr.12: 1027829-1027829 on Build GRCh38
rs140435811 Chr.12: 1027920-1027920 on Build GRCh38
rs527727581 Chr.12: 1027878-1027878 on Build GRCh38
rs772484675 Chr.12: 1028007-1028007 on Build GRCh38
rs759962535 Chr.12: 1027942-1027942 on Build GRCh38
rs771209499 Chr.12: 1028001-1028001 on Build GRCh38
rs562907468 Chr.12: 1027814-1027814 on Build GRCh38
rs111425264 Chr.12: 1027960-1027960 on Build GRCh38
rs371125113 Chr.12: 1027969-1027969 on Build GRCh38

View Full Product Details