Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 38309234-38309475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATL2
Entrez Gene ID
64225
Gene Name
atlastin GTPase 2
Gene Aliases
ARL3IP2, ARL6IP2, atlastin2
UniGene
Hs.594950
Gene Chromosome Location
Chr.2: 38293957-38378579 on Build GRCh38
Transcripts
NM_001135673.2 NM_001308076.1 NM_022374.3 NR_024191.1 XM_011533017.1 XM_011533018.2 XM_011533019.2 XM_011533020.1 XM_011533022.2 XM_011533023.2 XM_017004686.1 XM_017004687.1 XM_017004688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7112138 c.1049G>C p.R350T Chr.2: 38309401-38309401 on Build GRCh38
4313142 c.1051G>A p.D351N Chr.2: 38309399-38309399 on Build GRCh38
3183858 c.1051G>A p.D351N Chr.2: 38309399-38309399 on Build GRCh38
4943657 c.1047T>C p.C349C Chr.2: 38309403-38309403 on Build GRCh38
4664074 c.1020delA p.E341fs*2 Chr.2: 38309430-38309430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763101972 Chr.2: 38309436-38309436 on Build GRCh38
rs748907040 Chr.2: 38309335-38309335 on Build GRCh38
rs1002356624 Chr.2: 38309278-38309278 on Build GRCh38
rs770569074 Chr.2: 38309351-38309351 on Build GRCh38
rs778319950 Chr.2: 38309367-38309367 on Build GRCh38
rs766581581 Chr.2: 38309449-38309449 on Build GRCh38
rs1049941091 Chr.2: 38309274-38309276 on Build GRCh38
rs931566665 Chr.2: 38309273-38309273 on Build GRCh38
rs146778799 Chr.2: 38309290-38309290 on Build GRCh38
rs371444918 Chr.2: 38309450-38309450 on Build GRCh38
rs1010198562 Chr.2: 38309317-38309317 on Build GRCh38
rs771459737 Chr.2: 38309393-38309393 on Build GRCh38
rs968449924 Chr.2: 38309397-38309397 on Build GRCh38
rs762050463 Chr.2: 38309276-38309277 on Build GRCh38
rs773649649 Chr.2: 38309429-38309429 on Build GRCh38
rs200636844 Chr.2: 38309416-38309416 on Build GRCh38
rs777332951 Chr.2: 38309408-38309408 on Build GRCh38
rs1008213465 Chr.2: 38309309-38309309 on Build GRCh38
rs750250164 Chr.2: 38309359-38309360 on Build GRCh38
rs961000343 Chr.2: 38309297-38309297 on Build GRCh38
rs1035163887 Chr.2: 38309291-38309291 on Build GRCh38
rs79329134 Chr.2: 38309372-38309372 on Build GRCh38
rs1041428201 Chr.2: 38309359-38309359 on Build GRCh38
rs777482271 Chr.2: 38309331-38309331 on Build GRCh38
rs759722159 Chr.2: 38309451-38309451 on Build GRCh38

View Full Product Details