Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 50294679-50294918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY7
Entrez Gene ID
113
Gene Name
adenylate cyclase 7
Gene Aliases
AC7
UniGene
Hs.513578
Gene Chromosome Location
Chr.16: 50246158-50318135 on Build GRCh38
Transcripts
NM_001114.4 NM_001286057.1 XM_011522835.2 XM_011522836.2 XM_011522837.2 XM_011522838.2 XM_011522839.2 XM_011522840.2 XM_011522842.2 XM_017022895.1 XM_017022896.1 XM_017022897.1 XM_017022898.1 XM_017022899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6455628 c.891C>G p.P297P Chr.16: 50294694-50294694 on Build GRCh38
6866030 c.887C>A p.S296Y Chr.16: 50294690-50294690 on Build GRCh38
1493677 c.907G>C p.V303L Chr.16: 50294710-50294710 on Build GRCh38
1478910 c.943G>A p.A315T Chr.16: 50294746-50294746 on Build GRCh38
1158118 c.898C>A p.L300M Chr.16: 50294701-50294701 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376195841 Chr.16: 50294733-50294733 on Build GRCh38
rs1002811299 Chr.16: 50294857-50294857 on Build GRCh38
rs190808623 Chr.16: 50294894-50294894 on Build GRCh38
rs745987178 Chr.16: 50294767-50294768 on Build GRCh38
rs773974481 Chr.16: 50294736-50294736 on Build GRCh38
rs1011724122 Chr.16: 50294851-50294851 on Build GRCh38
rs758339490 Chr.16: 50294771-50294771 on Build GRCh38
rs1053314381 Chr.16: 50294821-50294821 on Build GRCh38
rs934906246 Chr.16: 50294804-50294804 on Build GRCh38
rs919720544 Chr.16: 50294762-50294762 on Build GRCh38
rs905645772 Chr.16: 50294856-50294856 on Build GRCh38
rs371452115 Chr.16: 50294756-50294756 on Build GRCh38
rs893293488 Chr.16: 50294836-50294836 on Build GRCh38
rs202146819 Chr.16: 50294743-50294743 on Build GRCh38
rs144754359 Chr.16: 50294788-50294788 on Build GRCh38
rs1040342801 Chr.16: 50294791-50294791 on Build GRCh38
rs928530955 Chr.16: 50294742-50294742 on Build GRCh38
rs753676126 Chr.16: 50294758-50294758 on Build GRCh38
rs770422373 Chr.16: 50294796-50294796 on Build GRCh38
rs376768521 Chr.16: 50294795-50294795 on Build GRCh38
rs772117582 Chr.16: 50294780-50294784 on Build GRCh38
rs752766398 Chr.16: 50294769-50294769 on Build GRCh38
rs767060124 Chr.16: 50294740-50294740 on Build GRCh38
rs577357633 Chr.16: 50294813-50294813 on Build GRCh38
rs760402594 Chr.16: 50294754-50294754 on Build GRCh38
rs538879220 Chr.16: 50294893-50294893 on Build GRCh38
rs761483130 Chr.16: 50294737-50294737 on Build GRCh38
rs1044620879 Chr.16: 50294854-50294854 on Build GRCh38
rs754851510 Chr.16: 50294852-50294852 on Build GRCh38
rs369936041 Chr.16: 50294760-50294760 on Build GRCh38
rs754889779 Chr.16: 50294759-50294759 on Build GRCh38
rs188700959 Chr.16: 50294787-50294787 on Build GRCh38
rs977862390 Chr.16: 50294746-50294746 on Build GRCh38
rs747150860 Chr.16: 50294780-50294780 on Build GRCh38

View Full Product Details