Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151218975-151219248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCF2
Entrez Gene ID
10061
Gene Name
ATP binding cassette subfamily F member 2
Gene Aliases
ABC28, EST133090, HUSSY-18, HUSSY18
UniGene
Hs.654958
Gene Chromosome Location
Chr.7: 151207837-151227374 on Build GRCh38
Transcripts
NM_005692.4 NM_007189.2 XM_017011639.1 XM_017011640.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3762505 c.939C>T p.Y313Y Chr.7: 151219142-151219142 on Build GRCh38
4816094 c.997G>C p.D333H Chr.7: 151219084-151219084 on Build GRCh38
4635488 c.980G>T p.R327M Chr.7: 151219101-151219101 on Build GRCh38
1488379 c.997G>C p.D333H Chr.7: 151219084-151219084 on Build GRCh38
6590648 c.931G>A p.D311N Chr.7: 151219150-151219150 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961037329 Chr.7: 151219009-151219009 on Build GRCh38
rs756440289 Chr.7: 151219171-151219171 on Build GRCh38
rs756207417 Chr.7: 151219034-151219034 on Build GRCh38
rs570238805 Chr.7: 151219179-151219179 on Build GRCh38
rs750683361 Chr.7: 151219033-151219033 on Build GRCh38
rs796348615 Chr.7: 151219032-151219034 on Build GRCh38
rs140619173 Chr.7: 151219156-151219156 on Build GRCh38
rs779314803 Chr.7: 151219072-151219072 on Build GRCh38
rs370845003 Chr.7: 151219100-151219100 on Build GRCh38
rs754020026 Chr.7: 151219178-151219178 on Build GRCh38
rs765142469 Chr.7: 151219133-151219133 on Build GRCh38
rs113981192 Chr.7: 151219009-151219009 on Build GRCh38
rs781339512 Chr.7: 151219031-151219031 on Build GRCh38
rs199903037 Chr.7: 151219174-151219174 on Build GRCh38
rs772791221 Chr.7: 151219088-151219088 on Build GRCh38
rs780305889 Chr.7: 151219037-151219037 on Build GRCh38
rs759491541 Chr.7: 151219132-151219132 on Build GRCh38
rs773629266 Chr.7: 151219028-151219030 on Build GRCh38
rs769084929 Chr.7: 151219069-151219069 on Build GRCh38
rs1055642995 Chr.7: 151219201-151219201 on Build GRCh38
rs142063362 Chr.7: 151219131-151219131 on Build GRCh38
rs893375084 Chr.7: 151219222-151219222 on Build GRCh38
rs758641731 Chr.7: 151219196-151219196 on Build GRCh38
rs911087215 Chr.7: 151219076-151219076 on Build GRCh38
rs919053007 Chr.7: 151219002-151219002 on Build GRCh38
rs770388025 Chr.7: 151219127-151219127 on Build GRCh38
rs752479304 Chr.7: 151219013-151219013 on Build GRCh38
rs748701263 Chr.7: 151219195-151219195 on Build GRCh38
rs376022395 Chr.7: 151219172-151219172 on Build GRCh38
rs972614397 Chr.7: 151219016-151219016 on Build GRCh38
rs904064656 Chr.7: 151219182-151219182 on Build GRCh38
rs985305619 Chr.7: 151219060-151219060 on Build GRCh38
rs182915092 Chr.7: 151219017-151219017 on Build GRCh38
rs6949587 Chr.7: 151219142-151219142 on Build GRCh38
rs749774035 Chr.7: 151219045-151219045 on Build GRCh38
rs760342842 Chr.7: 151219121-151219121 on Build GRCh38
rs200266285 Chr.7: 151219033-151219035 on Build GRCh38
rs1035314747 Chr.7: 151219006-151219006 on Build GRCh38
rs572437469 Chr.7: 151219184-151219184 on Build GRCh38
rs140336738 Chr.7: 151219073-151219073 on Build GRCh38
rs936828652 Chr.7: 151219191-151219191 on Build GRCh38
rs35656219 Chr.7: 151219029-151219031 on Build GRCh38
rs778141534 Chr.7: 151219208-151219208 on Build GRCh38
rs762591830 Chr.7: 151219150-151219150 on Build GRCh38
rs923247421 Chr.7: 151219141-151219141 on Build GRCh38
rs372499058 Chr.7: 151219170-151219170 on Build GRCh38
rs772488114 Chr.7: 151219081-151219081 on Build GRCh38
rs943419814 Chr.7: 151219123-151219123 on Build GRCh38

View Full Product Details