Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88643350-88643597 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYBA
Entrez Gene ID
1535
Gene Name
cytochrome b-245 alpha chain
Gene Aliases
p22-PHOX
UniGene
Hs.513803
Gene Chromosome Location
Chr.16: 88643289-88651084 on Build GRCh38
Transcripts
NM_000101.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6208375 c.421C>T p.R141W Chr.16: 88643520-88643520 on Build GRCh38
7321968 c.471G>T p.P157P Chr.16: 88643470-88643470 on Build GRCh38
6019233 c.534A>G p.G178G Chr.16: 88643407-88643407 on Build GRCh38
3749388 c.521T>C p.V174A Chr.16: 88643420-88643420 on Build GRCh38
7326153 c.452C>T p.P151L Chr.16: 88643489-88643489 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562308643 Chr.16: 88643546-88643546 on Build GRCh38
rs759771041 Chr.16: 88643523-88643523 on Build GRCh38
rs756973782 Chr.16: 88643453-88643453 on Build GRCh38
rs983923488 Chr.16: 88643491-88643491 on Build GRCh38
rs989507329 Chr.16: 88643512-88643512 on Build GRCh38
rs759482101 Chr.16: 88643417-88643417 on Build GRCh38
rs909600062 Chr.16: 88643516-88643516 on Build GRCh38
rs778207143 Chr.16: 88643576-88643576 on Build GRCh38
rs376631447 Chr.16: 88643458-88643458 on Build GRCh38
rs909652214 Chr.16: 88643391-88643391 on Build GRCh38
rs560810774 Chr.16: 88643575-88643575 on Build GRCh38
rs777768173 Chr.16: 88643430-88643430 on Build GRCh38
rs119103269 Chr.16: 88643568-88643568 on Build GRCh38
rs982199940 Chr.16: 88643533-88643533 on Build GRCh38
rs144748313 Chr.16: 88643407-88643408 on Build GRCh38
rs773556935 Chr.16: 88643415-88643415 on Build GRCh38
rs113033082 Chr.16: 88643488-88643488 on Build GRCh38
rs771202971 Chr.16: 88643515-88643515 on Build GRCh38
rs576565258 Chr.16: 88643508-88643508 on Build GRCh38
rs758404796 Chr.16: 88643423-88643423 on Build GRCh38
rs779612331 Chr.16: 88643480-88643480 on Build GRCh38
rs939664876 Chr.16: 88643393-88643393 on Build GRCh38
rs765297587 Chr.16: 88643530-88643530 on Build GRCh38
rs1043237243 Chr.16: 88643559-88643559 on Build GRCh38
rs72547285 Chr.16: 88643557-88643557 on Build GRCh38
rs748716081 Chr.16: 88643489-88643489 on Build GRCh38
rs898809017 Chr.16: 88643443-88643443 on Build GRCh38
rs1055333659 Chr.16: 88643440-88643440 on Build GRCh38
rs766523764 Chr.16: 88643416-88643416 on Build GRCh38
rs933701832 Chr.16: 88643545-88643545 on Build GRCh38
rs745307862 Chr.16: 88643407-88643407 on Build GRCh38
rs758709616 Chr.16: 88643542-88643542 on Build GRCh38
rs777676283 Chr.16: 88643367-88643367 on Build GRCh38
rs558222148 Chr.16: 88643466-88643466 on Build GRCh38
rs179363894 Chr.16: 88643570-88643570 on Build GRCh38
rs13306297 Chr.16: 88643468-88643468 on Build GRCh38
rs114610092 Chr.16: 88643538-88643538 on Build GRCh38
rs889390969 Chr.16: 88643547-88643547 on Build GRCh38
rs760991073 Chr.16: 88643509-88643509 on Build GRCh38
rs755779683 Chr.16: 88643561-88643561 on Build GRCh38
rs972548169 Chr.16: 88643526-88643526 on Build GRCh38
rs964575579 Chr.16: 88643473-88643473 on Build GRCh38
rs1031784335 Chr.16: 88643574-88643574 on Build GRCh38
rs780661682 Chr.16: 88643457-88643457 on Build GRCh38
rs1021677280 Chr.16: 88643446-88643446 on Build GRCh38
rs72667005 Chr.16: 88643429-88643429 on Build GRCh38
rs747488013 Chr.16: 88643578-88643578 on Build GRCh38
rs772514836 Chr.16: 88643501-88643501 on Build GRCh38
rs772492771 Chr.16: 88643414-88643414 on Build GRCh38
rs992691010 Chr.16: 88643470-88643470 on Build GRCh38
rs572654914 Chr.16: 88643436-88643436 on Build GRCh38
rs72547284 Chr.16: 88643461-88643461 on Build GRCh38
rs753554324 Chr.16: 88643418-88643420 on Build GRCh38
rs966768758 Chr.16: 88643532-88643532 on Build GRCh38
rs914005525 Chr.16: 88643408-88643408 on Build GRCh38
rs752613883 Chr.16: 88643422-88643422 on Build GRCh38
rs751430224 Chr.16: 88643551-88643551 on Build GRCh38
rs769229409 Chr.16: 88643410-88643410 on Build GRCh38
rs12123 Chr.16: 88643560-88643560 on Build GRCh38
rs760275837 Chr.16: 88643411-88643412 on Build GRCh38
rs889276822 Chr.16: 88643427-88643427 on Build GRCh38
rs1022849964 Chr.16: 88643471-88643471 on Build GRCh38
rs751340079 Chr.16: 88643451-88643451 on Build GRCh38
rs1008699438 Chr.16: 88643469-88643469 on Build GRCh38
rs104894515 Chr.16: 88643474-88643474 on Build GRCh38
rs774771301 Chr.16: 88643412-88643412 on Build GRCh38
rs763616635 Chr.16: 88643415-88643420 on Build GRCh38
rs779447060 Chr.16: 88643569-88643569 on Build GRCh38
rs746724477 Chr.16: 88643375-88643375 on Build GRCh38
rs904612575 Chr.16: 88643450-88643450 on Build GRCh38
rs776216387 Chr.16: 88643377-88643377 on Build GRCh38
rs776763356 Chr.16: 88643520-88643520 on Build GRCh38

View Full Product Details