Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15892252-15892511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4F2
Entrez Gene ID
8529
Gene Name
cytochrome P450 family 4 subfamily F member 2
Gene Aliases
CPF2
UniGene
Hs.558423
Gene Chromosome Location
Chr.19: 15878024-15898120 on Build GRCh38
Transcripts
NM_001082.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6084333 c.505G>C p.E169Q Chr.19: 15892329-15892329 on Build GRCh38
36627 c.446_447GG>AA p.R149Q Chr.19: 15892387-15892388 on Build GRCh38
4075125 c.456G>A p.T152T Chr.19: 15892378-15892378 on Build GRCh38
7363563 c.398G>C p.G133A Chr.19: 15892436-15892436 on Build GRCh38
3822198 c.480G>T p.L160L Chr.19: 15892354-15892354 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534638070 Chr.19: 15892309-15892309 on Build GRCh38
rs748370664 Chr.19: 15892484-15892484 on Build GRCh38
rs373698661 Chr.19: 15892282-15892282 on Build GRCh38
rs556151888 Chr.19: 15892398-15892398 on Build GRCh38
rs375526667 Chr.19: 15892355-15892355 on Build GRCh38
rs762812527 Chr.19: 15892319-15892319 on Build GRCh38
rs116286361 Chr.19: 15892453-15892453 on Build GRCh38
rs138811366 Chr.19: 15892352-15892352 on Build GRCh38
rs760735211 Chr.19: 15892420-15892420 on Build GRCh38
rs756442206 Chr.19: 15892275-15892275 on Build GRCh38
rs539029441 Chr.19: 15892439-15892439 on Build GRCh38
rs749173384 Chr.19: 15892484-15892485 on Build GRCh38
rs922678635 Chr.19: 15892360-15892360 on Build GRCh38
rs769787930 Chr.19: 15892392-15892392 on Build GRCh38
rs2906878 Chr.19: 15892470-15892470 on Build GRCh38
rs767627290 Chr.19: 15892346-15892346 on Build GRCh38
rs780906704 Chr.19: 15892389-15892389 on Build GRCh38
rs749206959 Chr.19: 15892476-15892476 on Build GRCh38
rs200629062 Chr.19: 15892379-15892379 on Build GRCh38
rs866778013 Chr.19: 15892357-15892357 on Build GRCh38
rs748349858 Chr.19: 15892408-15892408 on Build GRCh38
rs756904293 Chr.19: 15892456-15892456 on Build GRCh38
rs754089074 Chr.19: 15892373-15892373 on Build GRCh38
rs751186319 Chr.19: 15892455-15892455 on Build GRCh38
rs370785925 Chr.19: 15892378-15892378 on Build GRCh38
rs773403897 Chr.19: 15892489-15892489 on Build GRCh38
rs267605327 Chr.19: 15892387-15892387 on Build GRCh38
rs772148375 Chr.19: 15892409-15892409 on Build GRCh38
rs750011060 Chr.19: 15892462-15892462 on Build GRCh38
rs369799609 Chr.19: 15892279-15892279 on Build GRCh38
rs750911004 Chr.19: 15892351-15892351 on Build GRCh38
rs2906877 Chr.19: 15892457-15892457 on Build GRCh38
rs376639886 Chr.19: 15892276-15892276 on Build GRCh38
rs780136333 Chr.19: 15892438-15892439 on Build GRCh38
rs371325087 Chr.19: 15892391-15892391 on Build GRCh38
rs112646487 Chr.19: 15892415-15892415 on Build GRCh38
rs140630977 Chr.19: 15892388-15892388 on Build GRCh38
rs755662243 Chr.19: 15892463-15892463 on Build GRCh38
rs1014402937 Chr.19: 15892434-15892434 on Build GRCh38
rs2906875 Chr.19: 15892293-15892293 on Build GRCh38
rs61755872 Chr.19: 15892397-15892397 on Build GRCh38
rs766924159 Chr.19: 15892425-15892425 on Build GRCh38
rs576410035 Chr.19: 15892411-15892411 on Build GRCh38
rs770503691 Chr.19: 15892294-15892294 on Build GRCh38
rs777167655 Chr.19: 15892429-15892429 on Build GRCh38
rs367909140 Chr.19: 15892393-15892393 on Build GRCh38
rs779895249 Chr.19: 15892474-15892474 on Build GRCh38
rs769698813 Chr.19: 15892314-15892314 on Build GRCh38
rs902933201 Chr.19: 15892485-15892485 on Build GRCh38
rs746392149 Chr.19: 15892288-15892288 on Build GRCh38
rs374707045 Chr.19: 15892423-15892423 on Build GRCh38
rs772342427 Chr.19: 15892486-15892486 on Build GRCh38
rs774408246 Chr.19: 15892402-15892402 on Build GRCh38
rs761760699 Chr.19: 15892343-15892343 on Build GRCh38
rs144945582 Chr.19: 15892308-15892308 on Build GRCh38
rs778708764 Chr.19: 15892478-15892478 on Build GRCh38
rs2906876 Chr.19: 15892450-15892450 on Build GRCh38
rs774658023 Chr.19: 15892332-15892332 on Build GRCh38
rs775493462 Chr.19: 15892315-15892315 on Build GRCh38
rs568177101 Chr.19: 15892469-15892469 on Build GRCh38
rs767274275 Chr.19: 15892458-15892458 on Build GRCh38
rs768704426 Chr.19: 15892325-15892325 on Build GRCh38
rs754749070 Chr.19: 15892477-15892477 on Build GRCh38
rs911255628 Chr.19: 15892394-15892394 on Build GRCh38

View Full Product Details