Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 10972839-10973112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC16A
Entrez Gene ID
23274
Gene Name
C-type lectin domain family 16 member A
Gene Aliases
Gop-1, KIAA0350
UniGene
Hs.35490
Gene Chromosome Location
Chr.16: 10944488-11193272 on Build GRCh38
Transcripts
NM_001243403.1 NM_015226.2 XM_005255210.1 XM_005255211.1 XM_005255213.1 XM_005255214.1 XM_005255215.3 XM_005255216.1 XM_006720870.3 XM_011522434.1 XM_011522435.1 XM_011522436.2 XM_011522437.2 XM_011522438.2 XM_011522439.2 XM_011522440.2 XM_017023089.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3420745 c.629G>A p.R210Q Chr.16: 10972968-10972968 on Build GRCh38
2142011 c.605T>C p.L202S Chr.16: 10972938-10972938 on Build GRCh38
3420744 c.635G>A p.R212Q Chr.16: 10972968-10972968 on Build GRCh38
6423485 c.637A>T p.T213S Chr.16: 10972976-10972976 on Build GRCh38
5054620 c.706G>A p.V236M Chr.16: 10973045-10973045 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766392289 Chr.16: 10972910-10972910 on Build GRCh38
rs569055998 Chr.16: 10972993-10972993 on Build GRCh38
rs567868193 Chr.16: 10972867-10972868 on Build GRCh38
rs200325017 Chr.16: 10972907-10972907 on Build GRCh38
rs775473849 Chr.16: 10973084-10973084 on Build GRCh38
rs747501322 Chr.16: 10973029-10973029 on Build GRCh38
rs370420659 Chr.16: 10972902-10972902 on Build GRCh38
rs767288168 Chr.16: 10973074-10973074 on Build GRCh38
rs1021655085 Chr.16: 10972865-10972867 on Build GRCh38
rs761911979 Chr.16: 10972908-10972908 on Build GRCh38
rs776553109 Chr.16: 10973046-10973046 on Build GRCh38
rs1014789316 Chr.16: 10972910-10972910 on Build GRCh38
rs932287361 Chr.16: 10973081-10973081 on Build GRCh38
rs756837528 Chr.16: 10972991-10972991 on Build GRCh38
rs199587749 Chr.16: 10973086-10973086 on Build GRCh38
rs74697461 Chr.16: 10972924-10972924 on Build GRCh38
rs766250591 Chr.16: 10972909-10972909 on Build GRCh38
rs780647162 Chr.16: 10973023-10973023 on Build GRCh38
rs746434845 Chr.16: 10972882-10972882 on Build GRCh38
rs374378833 Chr.16: 10973069-10973069 on Build GRCh38
rs372457553 Chr.16: 10972958-10972958 on Build GRCh38
rs751645747 Chr.16: 10972909-10972910 on Build GRCh38
rs539958540 Chr.16: 10972884-10972884 on Build GRCh38
rs112472422 Chr.16: 10973073-10973073 on Build GRCh38
rs201136444 Chr.16: 10972945-10972945 on Build GRCh38
rs200859338 Chr.16: 10973064-10973064 on Build GRCh38
rs768899409 Chr.16: 10973043-10973043 on Build GRCh38
rs200841942 Chr.16: 10973087-10973087 on Build GRCh38
rs570750160 Chr.16: 10972881-10972881 on Build GRCh38
rs775739024 Chr.16: 10972887-10972887 on Build GRCh38
rs201921194 Chr.16: 10972889-10972889 on Build GRCh38
rs753566066 Chr.16: 10972967-10972967 on Build GRCh38
rs746191088 Chr.16: 10972999-10972999 on Build GRCh38
rs199556448 Chr.16: 10973059-10973059 on Build GRCh38
rs753299172 Chr.16: 10973036-10973046 on Build GRCh38
rs755474544 Chr.16: 10972915-10972916 on Build GRCh38
rs201475595 Chr.16: 10972959-10972959 on Build GRCh38
rs758974840 Chr.16: 10973006-10973006 on Build GRCh38
rs781574361 Chr.16: 10972923-10972923 on Build GRCh38
rs868304995 Chr.16: 10972950-10972950 on Build GRCh38
rs377250814 Chr.16: 10972955-10972955 on Build GRCh38

View Full Product Details