Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 65379323-65379596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC6
Entrez Gene ID
9829
Gene Name
DnaJ heat shock protein family (Hsp40) member C6
Gene Aliases
DJC6, PARK19
UniGene
Hs.647643
Gene Chromosome Location
Chr.1: 65264694-65415869 on Build GRCh38
Transcripts
NM_001256864.1 NM_001256865.1 NM_014787.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7309097 c.666+1G>T p.? Chr.1: 65379525-65379525 on Build GRCh38
6293786 c.412C>T p.L138L Chr.1: 65379441-65379441 on Build GRCh38
3491781 c.438G>A p.R146R Chr.1: 65379467-65379467 on Build GRCh38
5645803 c.608G>A p.R203Q Chr.1: 65379466-65379466 on Build GRCh38
7309096 c.666G>T p.L222F Chr.1: 65379524-65379524 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771207965 Chr.1: 65379366-65379366 on Build GRCh38
rs758360367 Chr.1: 65379346-65379346 on Build GRCh38
rs763024192 Chr.1: 65379376-65379376 on Build GRCh38
rs774761455 Chr.1: 65379513-65379513 on Build GRCh38
rs750201295 Chr.1: 65379387-65379387 on Build GRCh38
rs775893691 Chr.1: 65379562-65379567 on Build GRCh38
rs373591833 Chr.1: 65379534-65379534 on Build GRCh38
rs765297694 Chr.1: 65379358-65379358 on Build GRCh38
rs747941666 Chr.1: 65379558-65379558 on Build GRCh38
rs771322344 Chr.1: 65379467-65379467 on Build GRCh38
rs776854109 Chr.1: 65379457-65379457 on Build GRCh38
rs766346073 Chr.1: 65379527-65379527 on Build GRCh38
rs778346421 Chr.1: 65379422-65379422 on Build GRCh38
rs1054219624 Chr.1: 65379428-65379428 on Build GRCh38
rs760515930 Chr.1: 65379515-65379515 on Build GRCh38
rs766399234 Chr.1: 65379395-65379395 on Build GRCh38
rs752113204 Chr.1: 65379542-65379542 on Build GRCh38
rs753762106 Chr.1: 65379404-65379404 on Build GRCh38
rs759907494 Chr.1: 65379537-65379539 on Build GRCh38
rs549580287 Chr.1: 65379465-65379465 on Build GRCh38
rs757982943 Chr.1: 65379455-65379455 on Build GRCh38
rs770344233 Chr.1: 65379475-65379475 on Build GRCh38
rs777384906 Chr.1: 65379557-65379557 on Build GRCh38
rs774541369 Chr.1: 65379425-65379426 on Build GRCh38
rs777147830 Chr.1: 65379351-65379351 on Build GRCh38
rs974406930 Chr.1: 65379362-65379362 on Build GRCh38
rs775364521 Chr.1: 65379485-65379485 on Build GRCh38
rs768948356 Chr.1: 65379499-65379499 on Build GRCh38
rs746415427 Chr.1: 65379473-65379473 on Build GRCh38
rs780816481 Chr.1: 65379568-65379568 on Build GRCh38
rs2296480 Chr.1: 65379359-65379359 on Build GRCh38
rs781660857 Chr.1: 65379471-65379471 on Build GRCh38
rs1000848845 Chr.1: 65379403-65379403 on Build GRCh38
rs754507247 Chr.1: 65379417-65379417 on Build GRCh38
rs145570226 Chr.1: 65379357-65379357 on Build GRCh38
rs139223941 Chr.1: 65379502-65379502 on Build GRCh38
rs763124749 Chr.1: 65379487-65379487 on Build GRCh38
rs2296481 Chr.1: 65379393-65379393 on Build GRCh38
rs767803083 Chr.1: 65379544-65379546 on Build GRCh38
rs747328051 Chr.1: 65379466-65379466 on Build GRCh38
rs752269070 Chr.1: 65379437-65379437 on Build GRCh38
rs141323659 Chr.1: 65379382-65379382 on Build GRCh38
rs757927727 Chr.1: 65379543-65379543 on Build GRCh38

View Full Product Details