Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 105552755-105552996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF38
Entrez Gene ID
54848
Gene Name
Rho guanine nucleotide exchange factor 38
Gene Aliases
-
UniGene
Hs.585224
Gene Chromosome Location
Chr.4: 105552603-105682539 on Build GRCh38
Transcripts
NM_001242729.1 NM_017700.1 XM_011532050.2 XM_011532051.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2952857 c.175G>A p.E59K Chr.4: 105552940-105552940 on Build GRCh38
2952858 c.175G>A p.E59K Chr.4: 105552940-105552940 on Build GRCh38
5618760 c.156G>A p.R52R Chr.4: 105552921-105552921 on Build GRCh38
1642376 c.32A>C p.N11T Chr.4: 105552797-105552797 on Build GRCh38
6604311 c.39C>T p.V13V Chr.4: 105552804-105552804 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054558788 Chr.4: 105552933-105552933 on Build GRCh38
rs758535583 Chr.4: 105552846-105552846 on Build GRCh38
rs768788367 Chr.4: 105552902-105552902 on Build GRCh38
rs61753342 Chr.4: 105552826-105552826 on Build GRCh38
rs965565840 Chr.4: 105552780-105552780 on Build GRCh38
rs28672383 Chr.4: 105552942-105552942 on Build GRCh38
rs745919785 Chr.4: 105552786-105552786 on Build GRCh38
rs755727014 Chr.4: 105552829-105552829 on Build GRCh38
rs762338044 Chr.4: 105552816-105552816 on Build GRCh38
rs375048953 Chr.4: 105552948-105552948 on Build GRCh38
rs769002524 Chr.4: 105552812-105552812 on Build GRCh38
rs371638919 Chr.4: 105552860-105552860 on Build GRCh38
rs772317417 Chr.4: 105552791-105552791 on Build GRCh38
rs375204038 Chr.4: 105552818-105552818 on Build GRCh38
rs948661192 Chr.4: 105552851-105552851 on Build GRCh38
rs759047037 Chr.4: 105552821-105552821 on Build GRCh38
rs753565986 Chr.4: 105552956-105552956 on Build GRCh38
rs138889321 Chr.4: 105552841-105552841 on Build GRCh38
rs776862032 Chr.4: 105552815-105552815 on Build GRCh38
rs6533206 Chr.4: 105552939-105552939 on Build GRCh38
rs754240515 Chr.4: 105552873-105552874 on Build GRCh38
rs780289579 Chr.4: 105552878-105552878 on Build GRCh38
rs778960894 Chr.4: 105552843-105552843 on Build GRCh38
rs914387646 Chr.4: 105552937-105552937 on Build GRCh38
rs775849401 Chr.4: 105552808-105552808 on Build GRCh38
rs147662568 Chr.4: 105552824-105552824 on Build GRCh38
rs781528661 Chr.4: 105552904-105552904 on Build GRCh38
rs146139851 Chr.4: 105552834-105552834 on Build GRCh38
rs528842275 Chr.4: 105552925-105552925 on Build GRCh38
rs61413380 Chr.4: 105552823-105552823 on Build GRCh38
rs748432652 Chr.4: 105552906-105552906 on Build GRCh38
rs983609319 Chr.4: 105552833-105552833 on Build GRCh38
rs763464729 Chr.4: 105552930-105552930 on Build GRCh38
rs10032241 Chr.4: 105552921-105552921 on Build GRCh38
rs745866842 Chr.4: 105552844-105552844 on Build GRCh38
rs148016102 Chr.4: 105552940-105552940 on Build GRCh38
rs747288600 Chr.4: 105552811-105552811 on Build GRCh38
rs761508955 Chr.4: 105552957-105552957 on Build GRCh38
rs763912179 Chr.4: 105552831-105552831 on Build GRCh38
rs199819596 Chr.4: 105552886-105552886 on Build GRCh38
rs989839055 Chr.4: 105552835-105552835 on Build GRCh38

View Full Product Details