Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 26902475-26902706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH9
Entrez Gene ID
1007
Gene Name
cadherin 9
Gene Aliases
-
UniGene
Hs.272212
Gene Chromosome Location
Chr.5: 26880600-27038582 on Build GRCh38
Transcripts
NM_016279.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6866556 c.1151C>T p.T384I Chr.5: 26902578-26902578 on Build GRCh38
449510 c.1253A>G p.K418R Chr.5: 26902476-26902476 on Build GRCh38
279776 c.1037G>A p.R346K Chr.5: 26902692-26902692 on Build GRCh38
4671028 c.1165T>A p.L389M Chr.5: 26902564-26902564 on Build GRCh38
6546105 c.1088C>A p.P363H Chr.5: 26902641-26902641 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893053657 Chr.5: 26902635-26902635 on Build GRCh38
rs745991592 Chr.5: 26902662-26902662 on Build GRCh38
rs770856982 Chr.5: 26902561-26902561 on Build GRCh38
rs1055699145 Chr.5: 26902634-26902634 on Build GRCh38
rs755457072 Chr.5: 26902629-26902629 on Build GRCh38
rs867611896 Chr.5: 26902537-26902537 on Build GRCh38
rs909478234 Chr.5: 26902506-26902506 on Build GRCh38
rs144484970 Chr.5: 26902617-26902617 on Build GRCh38
rs774520401 Chr.5: 26902661-26902661 on Build GRCh38
rs182057099 Chr.5: 26902636-26902636 on Build GRCh38
rs373814780 Chr.5: 26902628-26902628 on Build GRCh38
rs866200603 Chr.5: 26902549-26902549 on Build GRCh38
rs139383777 Chr.5: 26902571-26902571 on Build GRCh38
rs376720983 Chr.5: 26902660-26902660 on Build GRCh38
rs146644878 Chr.5: 26902667-26902667 on Build GRCh38
rs767582762 Chr.5: 26902570-26902570 on Build GRCh38
rs759697917 Chr.5: 26902591-26902591 on Build GRCh38
rs974267404 Chr.5: 26902535-26902535 on Build GRCh38
rs748540104 Chr.5: 26902647-26902647 on Build GRCh38
rs756492421 Chr.5: 26902648-26902648 on Build GRCh38
rs757051985 Chr.5: 26902651-26902651 on Build GRCh38
rs753378587 Chr.5: 26902599-26902599 on Build GRCh38
rs973722890 Chr.5: 26902532-26902532 on Build GRCh38
rs941024304 Chr.5: 26902518-26902518 on Build GRCh38
rs775630425 Chr.5: 26902669-26902669 on Build GRCh38
rs756942748 Chr.5: 26902604-26902604 on Build GRCh38
rs760457024 Chr.5: 26902579-26902579 on Build GRCh38
rs150876106 Chr.5: 26902505-26902505 on Build GRCh38
rs774573447 Chr.5: 26902563-26902563 on Build GRCh38
rs778213959 Chr.5: 26902654-26902654 on Build GRCh38
rs568317141 Chr.5: 26902533-26902533 on Build GRCh38
rs749402733 Chr.5: 26902659-26902659 on Build GRCh38
rs929731772 Chr.5: 26902566-26902566 on Build GRCh38
rs748562835 Chr.5: 26902504-26902504 on Build GRCh38
rs201926799 Chr.5: 26902565-26902565 on Build GRCh38
rs773371878 Chr.5: 26902523-26902523 on Build GRCh38

View Full Product Details