Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104772891-104773134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKT1
Entrez Gene ID
207
Gene Name
AKT serine/threonine kinase 1
Gene Aliases
AKT, CWS6, PKB, PKB-ALPHA, PRKBA, RAC, RAC-ALPHA
UniGene
Hs.525622
Gene Chromosome Location
Chr.14: 104769349-104795743 on Build GRCh38
Transcripts
NM_001014431.1 NM_001014432.1 NM_005163.2 XM_005267401.1 XM_017021075.1 XM_017021076.1 XM_017021077.1 XM_017021078.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5044339 c.1069C>T p.L357F Chr.14: 104772981-104772981 on Build GRCh38
6981608 c.1061A>T p.H354L Chr.14: 104772989-104772989 on Build GRCh38
6929635 c.958G>T p.V320L Chr.14: 104773092-104773092 on Build GRCh38
6979438 c.1106C>T p.P369L Chr.14: 104772944-104772944 on Build GRCh38
5044337 c.1042C>T p.P348S Chr.14: 104773008-104773008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372502847 Chr.14: 104772963-104772963 on Build GRCh38
rs751709437 Chr.14: 104773063-104773063 on Build GRCh38
rs976500042 Chr.14: 104772999-104772999 on Build GRCh38
rs369048965 Chr.14: 104773099-104773099 on Build GRCh38
rs199770031 Chr.14: 104773098-104773098 on Build GRCh38
rs144088506 Chr.14: 104773030-104773030 on Build GRCh38
rs373253729 Chr.14: 104772927-104772927 on Build GRCh38
rs781749630 Chr.14: 104773077-104773077 on Build GRCh38
rs889182971 Chr.14: 104772932-104772932 on Build GRCh38
rs768409564 Chr.14: 104773105-104773105 on Build GRCh38
rs781388586 Chr.14: 104772938-104772938 on Build GRCh38
rs773975370 Chr.14: 104772958-104772958 on Build GRCh38
rs55874341 Chr.14: 104773072-104773072 on Build GRCh38
rs11555432 Chr.14: 104772980-104772980 on Build GRCh38
rs150399268 Chr.14: 104772943-104772943 on Build GRCh38
rs376942361 Chr.14: 104772997-104772997 on Build GRCh38
rs777711698 Chr.14: 104773060-104773060 on Build GRCh38
rs369520527 Chr.14: 104772950-104772950 on Build GRCh38
rs767544462 Chr.14: 104772991-104772991 on Build GRCh38
rs746272761 Chr.14: 104772941-104772941 on Build GRCh38
rs746397537 Chr.14: 104773079-104773079 on Build GRCh38
rs56289559 Chr.14: 104773018-104773018 on Build GRCh38
rs369527283 Chr.14: 104772916-104772916 on Build GRCh38
rs1050565251 Chr.14: 104772924-104772924 on Build GRCh38
rs758651475 Chr.14: 104773045-104773045 on Build GRCh38
rs1043166019 Chr.14: 104773039-104773039 on Build GRCh38
rs761646283 Chr.14: 104772988-104772988 on Build GRCh38
rs760425021 Chr.14: 104773000-104773000 on Build GRCh38
rs536727818 Chr.14: 104773108-104773108 on Build GRCh38
rs748884296 Chr.14: 104773100-104773100 on Build GRCh38
rs747408700 Chr.14: 104772937-104772937 on Build GRCh38
rs747738213 Chr.14: 104773111-104773111 on Build GRCh38
rs549370342 Chr.14: 104772942-104772942 on Build GRCh38
rs374865919 Chr.14: 104772928-104772928 on Build GRCh38

View Full Product Details