Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 103049616-103049889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNNM2
Entrez Gene ID
54805
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 2
Gene Aliases
ACDP2, HOMG6, HOMGSMR
UniGene
Hs.643509
Gene Chromosome Location
Chr.10: 102918293-103087169 on Build GRCh38
Transcripts
NM_017649.4 NM_199076.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
914290 c.1704C>T p.T568T Chr.10: 103049789-103049789 on Build GRCh38
914291 c.1720G>T p.E574* Chr.10: 103049805-103049805 on Build GRCh38
3433820 c.1703C>T p.T568I Chr.10: 103049788-103049788 on Build GRCh38
7313485 c.1750G>T p.E584* Chr.10: 103049835-103049835 on Build GRCh38
914292 c.1754C>T p.T585I Chr.10: 103049839-103049839 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752861468 Chr.10: 103049816-103049816 on Build GRCh38
rs759850444 Chr.10: 103049672-103049672 on Build GRCh38
rs387906975 Chr.10: 103049788-103049788 on Build GRCh38
rs925167429 Chr.10: 103049775-103049775 on Build GRCh38
rs151193266 Chr.10: 103049693-103049693 on Build GRCh38
rs754762020 Chr.10: 103049813-103049813 on Build GRCh38
rs766930033 Chr.10: 103049703-103049703 on Build GRCh38
rs7093326 Chr.10: 103049822-103049822 on Build GRCh38
rs371434890 Chr.10: 103049783-103049783 on Build GRCh38
rs770144844 Chr.10: 103049677-103049677 on Build GRCh38
rs893388419 Chr.10: 103049848-103049848 on Build GRCh38
rs781608944 Chr.10: 103049857-103049857 on Build GRCh38
rs777695456 Chr.10: 103049847-103049847 on Build GRCh38
rs1043614377 Chr.10: 103049727-103049727 on Build GRCh38
rs770130635 Chr.10: 103049862-103049862 on Build GRCh38
rs377640967 Chr.10: 103049743-103049743 on Build GRCh38
rs752121344 Chr.10: 103049672-103049672 on Build GRCh38
rs747058788 Chr.10: 103049849-103049849 on Build GRCh38
rs74464353 Chr.10: 103049701-103049701 on Build GRCh38
rs901937775 Chr.10: 103049861-103049861 on Build GRCh38
rs746383933 Chr.10: 103049859-103049859 on Build GRCh38
rs773817513 Chr.10: 103049680-103049680 on Build GRCh38
rs373729846 Chr.10: 103049734-103049734 on Build GRCh38
rs758121261 Chr.10: 103049688-103049689 on Build GRCh38
rs749987277 Chr.10: 103049718-103049718 on Build GRCh38

View Full Product Details